Skip to Content
Merck
All Photos(2)

Documents

339164

Sigma-Aldrich

Bis(cyclopentadienyl)cobalt(II)

Synonym(s):

Cobaltocene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(C5H5)2
CAS Number:
Molecular Weight:
189.12
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder or crystals
solid

reaction suitability

core: cobalt

mp

176-180 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[Co].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.Co/c2*1-2-4-5-3-1;/h2*1-5H;

InChI key

PXFGMRZPRDJDEK-UHFFFAOYSA-N

General description

Bis(cyclopentadienyl)cobalt(II) also known as cobaltocene, is an organometalliccompound that is widely used in the field of polymer synthesis, cobaltnanomaterials, and redox flow batteries.

Application

Bis(cyclopentadienyl)cobalt(II) can be used:
  • As a dopant to prepare encapsulated carbon nanotubes with high thermoelectric conversion efficiency.
  • As a CVD precursor to fabricate cobalt oxide thin films for various applications.
  • As a redox-active anode species in Li-based redox flow batteries to achieve higher energy densities and energy efficiencies.
  • As a catalyst for controlled/“living” radical polymerization of methylmethacrylate.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Sol. 2 - Muta. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A high-performance all-metallocene-based, non-aqueous redox flow battery
Yu Ding, et al.
Energy & Environmental Science, 10, 491-497 (2017)
Thin films of cobalt oxide deposited on high aspect ratio supports by atomic layer deposition
Madeleine Diskus, et al
Chem. Vap. Deposition, 17, 135-140 (2011)
F Li et al.
Chemical science, 9(30), 6379-6389 (2018-10-13)
A series of NO-bound, iron-functionalized polyoxovanadate-alkoxide (FePOV-alkoxide) clusters have been synthesized, providing insight into the role of multimetallic constructs in the coordination and activation of a substrate. Upon exposure of the heterometallic cluster to NO, the vanadium-oxide metalloligand is oxidized
Development of n-type cobaltocene-encapsulated carbon nanotubes with remarkable thermoelectric property
Takahiro Fukumaru, et al.
Scientific Reports, 5, 7951-7951 (2015)
Jacob M Clary et al.
Nanotechnology, 31(17), 175703-175703 (2020-01-09)
Highly dispersed cobalt atoms were deposited on porous alumina particles using atomic layer deposition (ALD) with a CoCp2/H2 chemistry at approximately 7 wt%. H2 did not completely reduce the cyclopentadienyl organic ligands bound to deposited Co atoms at ALD reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service