Skip to Content
Merck
All Photos(1)

Documents

665118

Sigma-Aldrich

2-Amino-5-methoxybenzoic acid

97%

Synonym(s):

6-Amino-m-anisic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2N)C6H3(OCH3)CO2H
CAS Number:
Molecular Weight:
167.16
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

148-152 °C

application(s)

peptide synthesis

SMILES string

COc1ccc(N)c(c1)C(O)=O

InChI

1S/C8H9NO3/c1-12-5-2-3-7(9)6(4-5)8(10)11/h2-4H,9H2,1H3,(H,10,11)

InChI key

UMKSAURFQFUULT-UHFFFAOYSA-N

Application

2-Amino-5-methoxybenzoic acid is a general reagent used in the synthesis of substituted isoquinolinonaphthyridines, quinazolinones, imidazobenzodiazepines, pyridoquinazolones and polycyclic hexahydrobenzo[c]acridines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of substituted imidazobenzodiazepines as novel asthma treatments.
Jahan R, et al.
European Journal of Medicinal Chemistry, 126, 550-560 (2017)
Bismuth (III) Chloride Catalyzed Intramolecular Hetero-Diels?Alder Reaction: Access to cis-Fused Angular Hexahydrobenzo[c]acridines.
Sabitha G, et al.
Synthesis, 47(01), 124-128 (2015)
Biologically active heterocyclic hybrids based on quinazolinone, benzofuran and imidazolium moieties: synthesis, characterization, cytotoxic and antibacterial evaluation.
Asadi P, et al.
Chemistry and Biodiversity, 14(4), e1600411-e1600411 (2017)
Condensation of anthranilic acids with pyridines to furnish pyridoquinazolones via pyridine dearomatization.
Yang Y, et al.
Chemical Communications (Cambridge, England), 52(87), 12869-12872 (2016)
Asymmetric synthesis of isoquinolinonaphthyridines catalyzed by a chiral Br?nsted acid.
Li J, et al.
Organic & Biomolecular Chemistry, 15(31), 6474-6477 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service