Skip to Content
Merck
All Photos(1)

Key Documents

8.06910

Sigma-Aldrich

n-Octane

for synthesis

Synonym(s):

n-Octane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH3
CAS Number:
Molecular Weight:
114.23
MDL number:
UNSPSC Code:
12352001
EC Index Number:
203-892-1
NACRES:
NA.22
grade:
for synthesis
bp:
125-126 °C/1013 hPa
vapor pressure:
14 hPa ( 20 °C)

grade

for synthesis

Quality Level

vapor pressure

14 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

210 °C

expl. lim.

0.8-6.5 % (v/v)

dilution

(for synthesis)

bp

125-126 °C/1013 hPa

mp

-57 °C

transition temp

flash point 13 °C

density

0.70 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCC)CCC

InChI

1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3

InChI key

TVMXDCGIABBOFY-UHFFFAOYSA-N

General description

n-octane is a hydrocarbon compound used as a solvent, cleaning agent, fuel additive, or reaction agents.

Application

n-Octane is used as:
  • A oil phase in the preparation of zinc carbonate particles by reverse microemulsion technique.
  • As a diluents in the solvent extraction of REEs(rare-earth elements) with extracting agent D2EHPA [di-(2-ethylhexyl)phosphoric acid].

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.702 - 0.704
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemistry for the generation of renewable chemicals: electrochemical conversion of levulinic acid
dos Santos, et al.
Royal Society of Chemistry Advances, 5(34), 26634-26643 (2015)
Synthesis of zinc carbonate hydroxide nanoparticles using microemulsion process
Alhawi T, et al.
Procedia Engineering, 102, 346-355 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service