Skip to Content
Merck
All Photos(1)

Key Documents

654892

Sigma-Aldrich

Aldrich® Negative Photoresist Kit I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

storage temp.

2-8°C

Quality Level

General description

Aldrich® Negative Photoresist Kit I consists of materials for photolithography. The photoresist can be spin coated on the glass substrate for the formation of electrode arrays. The kit also provides masking and patterning on the glass surface.

Application

Aldrich® Negative Photoresist Kit I has been used in the fabrication of microelectrodes by photolithography. It can also be used to fabricate the droplet-based microfluidic device, microfluidic chips, glass optical waveguides, and integrated grating couplers.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 651761Negative resist remover I 250 mLSDS

  • 651788Negative resist developer I 250 mLSDS

  • 651796Negative photoresist I 100 mLSDS

  • 651974Negative resist thinner I 100 mLSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication and characterization of optical waveguides and grating couplers
J P Sharpe, et al.
European Journal of Physics, 34, 1317-1317 (2013)
High-speed droplet actuation on single-plate electrode arrays
Banerjee AN, et al.
Journal of Colloid and Interface Science, 362(2), 567-574 (2011)
Arghya Narayan Banerjee et al.
Journal of colloid and interface science, 362(2), 567-574 (2011-08-02)
This paper reports a droplet-based microfluidic device composed of patterned co-planar electrodes in an all-in-a-single-plate arrangement and coated with dielectric layers for electrowetting-on-dielectric (EWOD) actuation of discrete droplets. The co-planar arrangement is preferred over conventional two-plate electrowetting devices because it
A compact 3D-printed interface for coupling open digital microchips with Venturi easy ambient sonic-spray ionization mass spectrometry
Hu J, et al.
Analyst, 140(5), 1495-1501 (2015)
Fabrication and characterization of optical waveguides and grating couplers
Sharpe JP, et al.
European Journal of Physics, 34(5), 1317-1317 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service