Skip to Content
Merck
All Photos(2)

Key Documents

651796

Sigma-Aldrich

Negative photoresist I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.23

mol wt

average Mw 60,000-70,000 (polyisoprene)

composition

solids, 27-29 wt. %

dielectric constant

2.4

surface tension

29.2 dyn/cm

viscosity

465-535 cP(25 °C)

bp

122-142 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

λmax

310-480 nm

storage temp.

2-8°C

SMILES string

O1C(OCC(C1)(C)C)CCCCCCCC

InChI

1S/C14H28O2/c1-4-5-6-7-8-9-10-13-15-11-14(2,3)12-16-13/h13H,4-12H2,1-3H3

InChI key

UBZVSDZJBLSIJG-UHFFFAOYSA-N

General description

A stabilized, purified photoresist which exhibits superior resolution, adhesion, etch resistance and low pinhole density.
Available as part of Negative Photoresist kit 654892

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gérald Guérin et al.
Journal of the American Chemical Society, 130(44), 14763-14771 (2008-10-14)
In alkane solvents, poly(isoprene-b-ferrocenyldimethylsilane) (PI-b-PFS) block copolymer forms fiberlike micelles, which show intriguing similarities with biological fibers such as amyloid fibers. Both systems exhibit fiber growth by a nucleated self-assembly mechanism and rapidly fragment upon exposure to the shear forces
Thomas Schmidt et al.
BMC biochemistry, 11, 11-11 (2010-02-23)
Natural rubber is a biopolymer with exceptional qualities that cannot be completely replaced using synthetic alternatives. Although several key enzymes in the rubber biosynthetic pathway have been isolated, mainly from plants such as Hevea brasiliensis, Ficus spec. and the desert
S Wołoszczuk et al.
The European physical journal. E, Soft matter, 33(4), 343-350 (2010-12-02)
We simulate ABA triblock copolymer melts using a lattice Monte Carlo method, known as cooperative motion algorithm, probing various degrees of compositional asymmetry. Selected order-disorder transition lines are determined in terms of the segment incompatibility, quantified by product χN
Stéphane Dubascoux et al.
Journal of chromatography. A, 1224, 27-34 (2012-01-17)
This paper presents results from the first analyses of the mesostructure of natural rubber (NR) by asymmetrical flow field flow fractionation (AF4). The results are compared with those obtained by size exclusion chromatography (SEC) in terms of average molar masses
S M Al-Salem et al.
Journal of hazardous materials, 172(2-3), 1690-1694 (2009-08-29)
Thermo-chemical treatments (mainly pyrolysis) directed towards energy and products recovery provide a very promising alternative to open space disposal or landfilling, reducing in the process hazardous waste and potential contamination to soil and water resources. In this communication, we present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service