Skip to Content
Merck
All Photos(1)

Documents

651818

Sigma-Aldrich

Gold etchant, standard

Synonym(s):

Antistrumin, Potassium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

composition

volatiles, 50-80%

bp

100 °C (lit.)
100 °C/1 atm

solubility

H2O: miscible

density

1.2924 g/mL at 25 °C (lit.)

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

General description

Etching readily controlled with minimum undercutting. Room temperature operated. Cyanide-free for standard applications. Etch rate of 28 angstrom/sec @ 25 °C.
Gold etchant is a combination of potassium iodide and iodine as complexing agents synthesized by electrochemically oxidizing gold.

Application

Gold etchant is a selective general-purpose etchant for gold, compatible with negative and positive photoresists. It is used to etch out gold (Au) electrodes prior to scanning electron microscopy (SEM) imaging for morphological study and metal layer for the preparation of nanoelectrodes array. Gold etchant can also be used in metal assisted chemical etching (MACE) for the fabrication of p-n junction silicon nanopillar based solar cells.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scalable long-term extraction of photosynthetic electrons by simple sandwiching of nanoelectrode array with densely-packed algal cell film.
Kim J et al.
Biosensors And Bioelectronics, 117(10), 15-22 (2018)
High performance nanopillars array silicon solar cells.
Design, Test, Integration and Packaging of MEMS/MOEMS (DTIP), 117(10), 1-5 (2013)
A two-step annealing process for enhancing the ferroelectric properties of poly (vinylidene fluoride)(PVDF) devices.
Park H, et al.
Journal of Material Chemistry C, 3(10), 2366-2370 (2015)
Cyanide-and thiourea-free electrochemical etching of gold for microelectronics applications.
Hu Z and Ritzdorf T
Journal of the Electrochemical Society, 154(10), D543-D549 (2007)
Abhinav Sharma et al.
Scientific reports, 9(1), 5970-5970 (2019-04-14)
Cardiac troponin T (cTnT) is considered a clinical standard for its high specificity and sensitivity when diagnosing acute myocardial infarction; however, most studies on the electrical sensors of cardiac troponin biomarkers have focused on cTnI rather than cTnT. This study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service