Skip to Content
Merck
All Photos(1)

Key Documents

649813

Sigma-Aldrich

PEDOT

tetramethacrylate end-capped solution, 0.5 wt. % (dispersion in propylene carbonate), contains p-toluenesulfonate as dopant

Synonym(s):

Oligotron PC, PEDOT, PEDOT, tetramethacrylate end-capped

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Product Name

Poly(3,4-ethylenedioxythiophene), tetramethacrylate end-capped solution, 0.5 wt. % (dispersion in propylene carbonate), contains p-toluenesulfonate as dopant

mol wt

~1,360-1,600 g/mol (methacrylate equivalent weight)
average Mn ~6,000(lit.)

Quality Level

contains

p-toluenesulfonate as dopant

composition

Oligotron tetramethacrylate, 0.5 wt. %
propylene carbonate, 99.5 wt. %

concentration

0.5 wt. % (dispersion in propylene carbonate)

resistance

1-10 M Ω/sq (typical surface resistance of film)

conductivity

0.1-0.5 S/cm (bulk)

density

1.189 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Please follow this link for Aedotron/Oligotron selection guide table.
Poly(3,4-ethylenedioxythiophene), tetramethacrylate end-capped solution (PEDOT-Meth) is a semiconductor based polymer that is functionalized with tetramethacrylate that facilitates effective crosslinking of the conjugated films.
The main characteristic of Oligotron PC is that it behaves as a reactive oligomer in non-volatile solvent. Used blended with monomers, or for reactions with functional compounds.

Application

For best results, agitate for 20 minutes prior to use. Films cast from propylene carbonate will require heat to dry. Photocurable inks can be formulated by adding acrylate monomers and photoinitiators (i.e. DMPA). Propylene carbonate will remain in the polymer. Functional compounds can be coupled to the oligomer via reactions at the methacrylate group.
PEDOT-Meth can be used as a plasticizing membrane in the fabrication of ion-selective sensors and light emitting diodes.
Product can be blended with additional acrylate or methacrylate monomers and crosslinked to make polymer blends. Custom molecules can be synthesized by bonding functional compounds, via the methacrylate groups, to the conducting EDOT oligomers.

Features and Benefits

End-capped structure renders the conducting oligomer PEDOT dispersible. The methacrylate groups offer the potential for crosslinking, or addtional chemical functionalization. Completely dried Oligotron(TM) tetramethacrylate via the methacrylate functional groups.

Caution

Store this product at room temperature (do not refrigerate or freeze). Some settling will normally occur. Agitate, sonicate, and filter prior to use. A plug of glass wool in a pipette can be used for easy filtration. The shelf life of this product will vary depending on how it is stored and handled. Product should be discarded if heavy aggregation occurs, or if sonication / filtering are inadequate to produce a good casting solution.

Legal Information

Oligotron is a trademark of TDA Research, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composite Polyacrylate- Poly (3, 4-ethylenedioxythiophene) Membranes for Improved All-Solid-State Ion-Selective Sensors
Rzewuska A, et al.
Analytical Chemistry, 80(1), 321-327 (2008)
Synthesis of water-free PEDOT with polyvinylpyrrolidone stabilizer in organic dispersant system
He J, et al.
Organic Electronics, 53, 117-126 (2018)
Cristina Ocaña et al.
Talanta, 186, 279-285 (2018-05-23)
We report here the fabrication of solid-contact calcium-selective electrodes (Ca2+-SCISEs) made of a polyurethane acrylate ion-selective membrane (ISM) that was covalently attached to the underlying ion-to-electron transducer (solid-contact). Methacrylate-functionalized poly(3,4-ethylenedioxythiophene) (Meth-PEDOT) and Meth-PEDOT films containing either multiwalled carbon nanotubes (MWCNT)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service