Skip to Content
Merck
All Photos(2)

Key Documents

678392

Sigma-Aldrich

PEDOT

nanotubes, hydrate

Synonym(s):

PEDOT, Poly(2,3-dihydrothieno-1,4-dioxin)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H4O2S)n ·xH2O
UNSPSC Code:
12352103
NACRES:
NA.23

product name

Poly(3,4-ethylenedioxythiophene) hydrate, nanotubes

form

nanotubes

L × I.D. × O.D.

~11.49 μm × 272.4 nm × 416 nm, TEM

General description

PEDOT is a conjugated, electronically and ionically active polymer. Conjugated polymers offer charge transport between inorganic, electrically conducting metals and organic, proton-conducting biological systems. PEDOT is chemically stable on account of the diethoxy substituents that replace the defect causing extra hydrogen atoms.The electron donating nature of the pendant PEDOT oxygen atoms also improves the conductivity of the molecule.

Application

Conjugated polymers such as PEDOT can be potentially useful for wide variety of bionic devices including cochlear, retinal, and cortical implants, as well as pacemakers and glucose sensors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang, X., et al.
Macromolecules, 39, 470-470 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service