641731
Titanium silicon oxide
nanopowder, <50 nm particle size (BET), 99.8% trace metals basis
Synonym(s):
Silicon titanate, Silicon titanium oxide, Titanium silicate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
Quality Level
Assay
99.8% trace metals basis
form
nanopowder
reaction suitability
reagent type: catalyst
core: titanium
particle size
<50 nm (BET)
density
3.39 g/mL at 25 °C (lit.)
SMILES string
[Ti+4].[O-][Si]([O-])([O-])[O-]
InChI
1S/O4Si.Ti/c1-5(2,3)4;/q-4;+4
InChI key
CFRNXBBHKHHBQM-UHFFFAOYSA-N
Other Notes
Amorphous mixture of SiO2 and TiO2.
Legal Information
Product of Engi-Mat Co.
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service