Skip to Content
Merck
All Photos(1)

Key Documents

594199

Sigma-Aldrich

Poly[(p-phenylenevinylene)-alt-(2-methoxy-5-(2-ethylhexyloxy)-p-phenylenevinylene)]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C25H32O2)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

Quality Level

solubility

chloroform: soluble
organic solvents: soluble

fluorescence

λex 451 nm; λem 517 nm in chloroform

General description

Poly[(p-phenylenevinylene)-alt-(2-methoxy-5-(2-ethylhexyloxy)-p-phenylenevinylene)] (p-PMEH-PPV) is a conducting polymer that can be used as an active material in the development of optoelectronic devices. It forms a conjugated-nonconjugated multiblock copolymer which can be used as an electroluminescent polymeric material. It can be potentially operated at low voltages with quick responsive and defined displays.

Application

p-PMEH-PPV can be used as a blue EL polymer for the fabrication of semiconductor devices such as organic light emitting diodes (OLEDs), thin film transistors (TFTs), photovoltaic cells and chemical sensors.

Features and Benefits

Light emission blue shifted relative to that for MEH-PPV by introduction of unsubstituted phenylene units with para linkage in the polymer main chain, shortening the effective conjugation length of the polymer.
Light emitting semiconjugated alternating copolymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and photo-induced charge separation of confined conjugation length phenylene vinylene-based polymers
Tan TAT, et al.
Polym. Chem., 4(20), 5305-5309 (2013)
High-efficiency poly (p-phenylenevinylene)-based copolymers containing an oxadiazole pendant group for light-emitting diodes
Jin S, et al.
Journal of the American Chemical Society, 126(8), 2474-2480 (2004)
Blue electroluminescent polymers: control of conjugation length by kink linkages and substituents in the poly (p-phenylenevinylene)-related copolymers
Ahn T, et al.
Macromolecules, 32(10), 3279-3285 (1999)
A molecular photosensor based on photoswitching of charge carrier mobility
Macromolecular Symposia, 247(1), 318-325 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service