Skip to Content
Merck
All Photos(1)

Key Documents

637262

Sigma-Aldrich

Titanium(IV) oxide, rutile

nanopowder, <100 nm particle size, 99.5% trace metals basis

Synonym(s):

TiO2 rutile, rutile titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.5% trace metals basis

form

nanopowder

diam. × L

~10 nm × 40 nm

surface area

50 m2/g

particle size

<100 nm

density

4.17 g/mL at 25 °C (lit.)

bulk density

0.06‑0.10 g/mL

application(s)

battery manufacturing

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rutile titanium(IV) oxide, also called titanium dioxide, is a fine powder with a particle size less than 100 nm. This titanium adopts the rutile crystal structure and is a white, opaque, crystalline solid with a high refractive index. It is widely used as a pigment in paints, plastics, paper, and cosmetics. Rutile titanium dioxide nanopowder has a high surface area, making it more reactive and effective in a range of applications. It is resistant to heat and chemical attack, making it suitable for use in high-temperature and corrosive environments.

Application

  • A study on titanium dioxide nanoparticles synthesized from titanium isopropoxide under SILAR-induced gel method: Transition from anatase to rutile structure: This research explores the synthesis and phase transition of titanium dioxide nanoparticles from anatase to rutile structure using the SILAR-induced gel method (AC Nkele et al., 2020).
  • Synthesis of rutile TiO2 nanostructures by single step hydrothermal route and its characterization: This article describes the synthesis of rutile TiO2 nanostructures using a single-step hydrothermal method and their characterization (SB Wategaonkar et al., 2020).
  • Monolayer Intermixed Oxide Surfaces: Fe, Ni, Cr, and V Oxides on Rutile TiO2(011): The study investigates the formation of mixed oxide layers on rutile TiO2(011) and their structural properties (S Halpegamage et al., 2016).
  • Mechanism, thermodynamics and kinetics of rutile leaching process by sulfuric acid reactions: This research examines the dissolution of rutile in sulfuric acid, focusing on the thermodynamics and kinetics of the process (AV Dubenko et al., 2020).
  • Kinetics of anatase transition to rutile TiO2 from titanium dioxide precursor powders synthesized by a sol-gel process: This paper studies the phase transition kinetics of anatase to rutile TiO2 from sol-gel synthesized precursor powders (CL Wang et al., 2016).

Features and Benefits

Possesses improved photocatalytic activity.

Other Notes

May contain up to 5 wt. % Silicon dioxide as a surface coating.

Contains small amount of anatase.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suxin Gui et al.
Journal of agricultural and food chemistry, 61(37), 8959-8968 (2013-08-24)
TiO₂ nanoparticles (NPs) are used in the food industry but have potential toxic effects in humans and animals. TiO₂ NPs impair renal function and cause oxidative stress and renal inflammation in mice, associated with inhibition of nuclear factor erythroid-2-related factor
D Minetto et al.
Environment international, 66, 18-27 (2014-02-11)
The innovative properties of nanomaterials make them suitable for various applications in many fields. In particular, TiO2 nanoparticles (nTiO2) are widely used in paints, in cosmetics and in sunscreens that are products accessible to the mass market. Despite the great
Roberta Tassinari et al.
Nanotoxicology, 8(6), 654-662 (2013-07-10)
The study explored possible reproductive and endocrine effects of short-term (5 days) oral exposure to anatase TiO2 nanoparticles (0, 1, 2 mg/kg body weight per day) in rat. Nanoparticles were characterised by scanning electron microscopy (SEM) and transmission electron microscopy
Nabila Haddou et al.
Chemosphere, 107, 304-310 (2014-01-28)
The Gliding Arc Discharge (GAD) is an efficient non-thermal plasma technique able to degrade organic compounds dispersed in water at atmospheric pressure. The degradation of the organometallic lead acetate (PbAc) in aqueous solution was performed by two distinct plasmageneous processes:
Elisa Moschini et al.
Toxicology letters, 222(2), 102-116 (2013-08-03)
Metal oxide NPs are abundantly produced in nanotech industries and are emitted in several combustion processes, suggesting the need to characterize their toxic impact on the human respiratory system. The acute toxicity and the morphological changes induced by copper oxide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service