634689
Strontium titanate
single crystal substrate, <100>
Synonym(s):
Strontium titanium trioxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
crystalline (cubic (a=3.905 Å))
dielectric constant
~300
hardness
6 (, Mohs)
reaction suitability
reagent type: catalyst
core: titanium
size
10 mm × 10 mm × 0.5 mm
mp
2060 °C (lit.)
2080 °C
density
4.81 g/mL at 25 °C (lit.)
5.175 g/mL at 25 °C
semiconductor properties
<100>
SMILES string
[Sr++].[O-][Ti]([O-])=O
InChI
1S/3O.Sr.Ti/q;2*-1;+2;
InChI key
VEALVRVVWBQVSL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Physical properties
Loss Tangent at 10GHz: ~5 x 10-4 @ 300K, ~3 x 10-4 @ 77K; Thermal expansion: 10.4 (x 10-6/°C)
Physical form
cubic (a = 3.905 Å)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nature nanotechnology, 6(6), 343-347 (2011-04-19)
Devices that confine and process single electrons represent an important scaling limit of electronics. Such devices have been realized in a variety of materials and exhibit remarkable electronic, optical and spintronic properties. Here, we use an atomic force microscope tip
Journal of the American Chemical Society, 134(4), 1974-1977 (2012-01-28)
A strategy of reaction-environment modulation was employed to change the surface property of a semiconductor photocatalyst to enhance its photocatalytic performance. Surface alkalinization induced by a high alkalinity of the solution environment significantly shifted the surface energy band of a
Polar liquid molecule induced transport property modulation at LaAlO₃/SrTiO₃ heterointerface.
Advanced materials (Deerfield Beach, Fla.), 24(19), 2598-2602 (2012-04-13)
Tailoring interface structure in highly strained YSZ/STO heterostructures.
Advanced materials (Deerfield Beach, Fla.), 23(44), 5268-5274 (2012-02-03)
Advanced materials (Deerfield Beach, Fla.), 24(21), 2885-2889 (2012-04-21)
A synthetic method of using genetically engineered M13 virus to mineralize perovskite nanomaterials, particularly strontium titanate (STO) and bismuth ferrite (BFO), is presented. Genetically engineered viruses provide effective templates for perovskite nanomaterials. The virus-templated nanocrystals are small in size, highly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service