Skip to Content
Merck
All Photos(1)

Key Documents

95420

Sigma-Aldrich

Tungstic acid

≥98.0% (calcined substance, T), powder

Synonym(s):

Tungstic(VI) acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2WO4
CAS Number:
Molecular Weight:
249.85
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥98.0% (calcined substance, T)

form

powder

impurities

≤0.2% insoluble matter in NaOH

loss

≤9.0% loss on ignition

density

5.5 g/mL at 25 °C (lit.)

SMILES string

O[W](O)(=O)=O

InChI

1S/2H2O.2O.W/h2*1H2;;;/q;;;;+2/p-2

InChI key

CMPGARWFYBADJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Tungstic acid may be prepared by either digesting tungstate of lime or by treating tungsten with hydrochloric acid. 1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P T Bilous et al.
Journal of bacteriology, 162(3), 1151-1155 (1985-06-01)
Escherichia coli grew anaerobically on a minimal medium with glycerol as the carbon and energy source and dimethyl sulfoxide (DMSO) as the terminal electron acceptor. DMSO reductase activity, measured with an artificial electron donor (reduced benzyl viologen), was preferentially associated
Chao-yang Tu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 25(6), 804-807 (2005-10-06)
Cr3+:Al2(WO4)3 Crystal was grown from the Na2WO4 solvent system by using the TSSG method. The absorption spectrum shows that there are two strong absorption wideband peaks and one we ak absorption peak belonging to Cr3+ ion in the crystal. The
Xiankun Lin et al.
Langmuir : the ACS journal of surfaces and colloids, 25(11), 6081-6087 (2009-03-12)
Hexagonally mesostructured polyoxometalate-based hybrid was prepared by a mild-solution method through the self-assembly of an amphiphilic poly(ethylene oxide) octadecyldimethylammonium (C18NEO12) and a polyanionic silicotungstic acid (HSiW). The composition of the hybrid was characterized through 1H NMR, infrared spectroscopy, mass spectroscopy
R R Bridges et al.
Journal of analytical toxicology, 8(6), 263-268 (1984-11-01)
Common techniques for analyzing ethchlorvynol (Placidyl) by colorimetry and gas liquid chromatography are evaluated. Matrix effects are thoroughly examined to determine their contribution to the validity of results.
Renagul Abdurahman et al.
Analytica chimica acta, 658(1), 63-67 (2010-01-20)
In this study, a sensitive optical waveguide (OWG) sensor for the detection and identification of volatile organic compounds (VOCs) was reported. The sensing membrane is constructed by immobilization of peroxopolytungsten acid (PTA) thin film over a single-mode potassium ion (K(+))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service