Merck
  • Biotemplated synthesis of perovskite nanomaterials for solar energy conversion.
Biotemplated synthesis of perovskite nanomaterials for solar energy conversion.
Advanced materials (Deerfield Beach, Fla.) (2012-04-21)
Nurxat Nuraje, Xiangnan Dang, Jifa Qi, Mark A Allen, Yu Lei, Angela M Belcher
ABSTRACT
A synthetic method of using genetically engineered M13 virus to mineralize perovskite nanomaterials, particularly strontium titanate (STO) and bismuth ferrite (BFO), is presented. Genetically engineered viruses provide effective templates for perovskite nanomaterials. The virus-templated nanocrystals are small in size, highly crystalline, and show photocatalytic and photovoltaic properties.
Sigma-Aldrich
Strontium titanate, single crystal substrate, <100>
Sigma-Aldrich
Calcium titanate, nanopowder, <100 nm particle size (BET), 99% trace metals basis
Sigma-Aldrich
Strontium titanate, nanopowder, <100 nm particle size, 99% trace metals basis
Sigma-Aldrich
Strontium titanate, powder, 99%