Skip to Content
Merck
All Photos(1)

Key Documents

634131

Sigma-Aldrich

Aluminum titanate

nanopowder, <25 nm particle size (BET), 98.5% trace metals basis

Synonym(s):

Aluminum titanium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al2TiO5
CAS Number:
Molecular Weight:
181.83
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98.5% trace metals basis

form

nanopowder

particle size

<25 nm (BET)

SMILES string

O=[Al]O[Ti](=O)O[Al]=O

InChI

1S/2Al.5O.Ti

InChI key

CNRZQDQNVUKEJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Material has high reactivity, low thermal conductivity, high thermal shock resistance, and low thermal expansion.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service