Skip to Content
Merck
All Photos(1)

Key Documents

62240

Sigma-Aldrich

Linoleic acid

technical, 58-74% (GC)

Synonym(s):

cis-9,cis-12-Octadecadienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
280.45
Beilstein:
1727101
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical

form

liquid

concentration

58-74% (GC)

impurities

18-32% oleic acid

color

clear

refractive index

n20/D 1.466 (lit.)
n20/D 1.466

bp

229-230 °C/16 mmHg (lit.)

mp

−5 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(CCCCCCC/C=C\C/C=C\CCCCC)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-

InChI key

OYHQOLUKZRVURQ-HZJYTTRNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Linoleic acid can give rise to a class of geometric, positional and conjugated dienoic isomers called conjugated linoleic acids(CLA)[1],mostly found in animal products[2][3], which can act like a cancer preventive agent in animals.[4]

Application

  • Alginate Spheres With Urea and Linoleic Acid: A biological nitrification inhibitor proposal utilizing alginate spheres to encapsulate urea and linoleic acid for agricultural benefits (Avendaño-Morales et al., 2024).

Biochem/physiol Actions

Linoleic acid increases cell proliferation and gene expression of PPARα and its target genes such as acyl-CoA oxidase in primary duck hepatocytes [5].

Caution

partially crystallizes on storage at 0-4°C

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Dietary fatty acid sources affect conjugated linoleic acid concentrations in milk from lactating dairy cows"
Kelly.LM, et al.
The Journal of Nutrition, 128(05), 881-885 (1998)
"Dietary trans-10, cis-12 conjugated linoleic acid induces hyperinsulinemia and fatty liver in the mouse"
Clement L, et al.
Journal of Lipid Research, 43(09), 1400-1409 (2002)
"Conjugated linoleic acid supplementation reduces adipose tissue by apoptosis and develops lipodystrophy in mice"
Tsuboyama-Kasaoka N, et al.
Diabetes, 49(09), 1534-1542 (2000)
"Conjugated linoleic acid?enriched butter fat alters mammary gland morphogenesis and reduces cancer risk in rats"
Ip C, et al.
The Journal of Nutrition, 129(12), 2135-2142 (1999)
Saman Lashkari et al.
Food science & nutrition, 8(1), 79-87 (2020-01-30)
Manipulation of rumen biohydrogenation (BH) is of great importance, since decreased BH of linolenic acid (LNA) and linoleic acid (LA) is linked to increased content of the beneficial polyunsaturated fatty acids (PUFA) in dairy products and decreased content of trans fatty

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service