575208
Lanthanum(III) sulfate
≥99.99% trace metals basis
Synonym(s):
Dilanthanum trisulfate, Lanthanum sulfate, Lanthanum sulfate (La2 (SO4)3)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
La2(SO4)3
CAS Number:
Molecular Weight:
566.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder
Recommended Products
Quality Level
Assay
≥99.99% trace metals basis
form
powder
reaction suitability
reagent type: catalyst
core: lanthanum
impurities
≤100.0 ppm Trace Rare Earth Analysis
SMILES string
[La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
InChI
1S/2La.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6
InChI key
VQEHIYWBGOJJDM-UHFFFAOYSA-H
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A C Blackburn et al.
Acta crystallographica. Section C, Crystal structure communications, 50 ( Pt 6), 833-838 (1994-06-15)
The structure of NaLaIII(SO4)2.H2O consists of distorted Na-O6 octahedra, La-O9 complexes in the form of distorted tricapped trigonal prisms, and slightly irregular tetrahedral sulfate ions. All bond distances fall within normal limits. The sulfate ions manifest rigid-body behavior but this
S Ranganayaki et al.
Zeitschrift fur allgemeine Mikrobiologie, 21(4), 329-332 (1981-01-01)
The effect of lanthanum sulphate on five Azotobacter strains (4A7, 10, 5A2, 5A1, and 3A3) isolated from Allahabad soil consists in a considerable increase in the nitrogen fixation, carbon consumption and in the amount of nitrogen fixed in mg per
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service