Skip to Content
Merck
All Photos(2)

Documents

359009

Sigma-Aldrich

Cerium(IV) sulfate

Synonym(s):

Ceric sulfate, Cerium disulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(SO4)2
CAS Number:
Molecular Weight:
332.24
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

core: cerium
reagent type: catalyst

density

3.01 g/mL at 25 °C (lit.)

SMILES string

[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Ce.2H2O4S/c;2*1-5(2,3)4/h;2*(H2,1,2,3,4)/q+4;;/p-4

InChI key

VZDYWEUILIUIDF-UHFFFAOYSA-J

Application

Cerium(IV) sulfate can be used to catalyze:
  • Selective oxidation of secondary alcohols to the corresponding ketones using sodium bromate as the oxidant.
  • Direct sulfonation of methane with sulfur trioxide to methanesulfonic acid in sulfuric acid.
  • Synthesis of 14-aryl- or 14-alkyl-14H-dibenzo[aj]xanthenes from the condensation of 2-naphthol and aldehydes under solvent-free and microwave conditions.

Reactant for:
  • Preparation of spherical-like nanoparticles CeO2 nanoparticles for NO reductions reactions

  • Oxidizing agent

Other Notes

may contain excess sulfate

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John H T Luong et al.
The journal of physical chemistry. B, 109(4), 1400-1407 (2006-07-21)
A simple wet chemical method involving only ultrasonic processing in dilute ceric sulfate (CS) was used to functionalize carbon nanotubes (CNTs). Unexpectedly, single-walled and multiwalled carbon nanotubes (SWCNTs and MWCNTs) were cut, oxidized, and disintegrated by sonication in 0.1 N
Toshio Yao et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 19(6), 829-833 (2003-07-02)
A flow injection biosensor system was proposed for the highly sensitive detection of 2,4,6-trichlorophenol (2,4,6-TCP). The system is based on the preoxidation by ceric sulfate to the corresponding benzoquinone (2,6-dichloro-1,4-benzoquinone: 2,6-DC-1,4-BQ), which was characterized using cyclic voltammetry, hydrodynamic voltammetry, and
A Porvali et al.
Waste management (New York, N.Y.), 107, 66-73 (2020-04-12)
Efficient recovery of REEs present in the battery waste is a modern problem that has proven to be difficult to solve in an efficient manner. The raw material investigated in the current study is mixed alkaline rare earth element (REE)
Hidayat Hussain et al.
Chemistry & biodiversity, 12(2), 289-294 (2015-02-14)
A new polychlorinated triphenyl diether named microsphaerol (1), has been isolated from the endophtic fungus Microsphaeropsis sp. An intensive phytochemical investigation of the endophytic fungus Seimatosporium sp., led to the isolation of a new naphthalene derivative named seimatorone (2) and
S M Sultan et al.
Journal of pharmaceutical and biomedical analysis, 7(3), 279-286 (1989-01-01)
A simple and accurate kinetic method for the determination of propranolol has been developed. Cerium(IV) sulphate (0.5 M) is used to oxidize propranolol in 2 M sulphuric acid at room temperature to the ketone form that absorbs light at a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service