Skip to Content
Merck
All Photos(1)

Documents

571245

Sigma-Aldrich

Pyrene

sublimed grade, 99%

Synonym(s):

Benzo[def]phenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
Beilstein:
1307225
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Quality Level

Assay

99%

mp

145-148 °C (lit.)

fluorescence

λex 338 nm; λem 375 nm in DMSO

SMILES string

c1cc2ccc3cccc4ccc(c1)c2c34

InChI

1S/C16H10/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h1-10H

InChI key

BBEAQIROQSPTKN-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

435.2 °F

Flash Point(C)

224 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chufen Yang et al.
Materials science & engineering. C, Materials for biological applications, 84, 254-262 (2018-03-10)
To improve the drug release rate in well-controlled manner, a new pH-sensitive triblock amphiphilic copolymer methyl poly(ethylene glycol) ether-b-poly(β-amino esters)-b-poly lactic acid (MPEG-b-PBAE-b-PLA) and its self-assembled micelles were developed for anticancer drug delivery. The average molecular weight and molecular structure
Tatsuya Itoyama et al.
Marine pollution bulletin, 124(2), 792-797 (2017-03-06)
Pyrene, a member of the polycyclic aromatic hydrocarbons (PAHs), contributes to abnormality in the size of the brain and the swimming behavior of pufferfish (Takifugu niphobles) larvae. We hypothesized that the aryl hydrocarbon receptor (AHR) may mediate pyrene-induced toxic effects
Yeimy J Rodriguez et al.
Pharmaceutics, 12(3) (2020-02-29)
Amphotericin B (AmB) is a broad spectrum of antifungal drug used to treat antifungal diseases. However, due to the high toxicity of AmB, treated patients may suffer the risk of side effects, such as renal failure. Nanoencapsulation strategies have been
Hao Wang et al.
Acta biomaterialia, 72, 206-216 (2018-03-24)
While polysaccharide-based nanocarriers have been recognized for their crucial roles in tumor theranostics, the industrial-scale production of nanotherapeutics still remains a significant challenge. Most current approaches adopt a postpolymerization self-assembly strategy that follows a separate synthetic step and thus suffers
Tomohiro Kato et al.
Journal of the American Chemical Society, 135(2), 741-750 (2012-12-18)
Although distance dependence of Förster resonance energy transfer (FRET) is well-studied and FRET has been extensively applied as "molecular ruler", only limited examples of orientation-dependent FRET have been reported. To create a robust FRET system that precisely reflects the orientation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service