Skip to Content
Merck
All Photos(1)

Documents

694959

Sigma-Aldrich

Anthracene

sublimed grade, ≥99%

Synonym(s):

Anthraxcene, Paranaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
Colour Index Number:
10790
Beilstein:
1905429
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39011608
PubChem Substance ID:
NACRES:
NA.22

grade

sublimed grade

Quality Level

vapor density

6.15 (vs air)

vapor pressure

1 mmHg ( 145 °C)

Assay

≥99%

form

crystalline

autoignition temp.

1004 °F

loss

0.5 wt. % loss on heating, 155°C (typical, TGA)

bp

340 °C (lit.)

mp

210-215 °C (lit.)

solubility

alcohols: soluble
benzene: soluble
chloroform: soluble
hydronaphthalenes: soluble
supercritical carbon dioxide: soluble

Orbital energy

HOMO 5.7 eV 
LUMO 1.7 eV 

SMILES string

c1ccc2cc3ccccc3cc2c1

InChI

1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H

InChI key

MWPLVEDNUUSJAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TGA/DSC Lot specific traces available upon request

Application

Anthracene has been shown to be soluble in a variety of binary and ternary mixtures of cyclohexanone, ethyl acetate, and methanol .

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Payam Payamyar et al.
Chimia, 67(4), 283-285 (2013-08-24)
We describe the challenges involved with extending the limited lateral size of two-dimensional polymers (2DPs). An amphiphilic monomer with three-fold symmetry is chosen to form an ideally tessellated monolayer at the air/water interface. Anthracene [4+4] photo-dimerization is chosen as the
Hyunjung Lee et al.
Inorganic chemistry, 51(20), 10904-10915 (2012-09-26)
The tendency of a Hg(II) ion to strongly quench fluorescence of potential fluorescent sensors is explored. Fluorescence measurements show the expected order of the chelation-enhanced fluorescence (CHEF) effect of Zn(II) > Cd(II) > Hg(II) ~ Cu(II), which is interpreted as
Alina P Sergeeva et al.
Journal of the American Chemical Society, 134(43), 18065-18073 (2012-10-04)
Clusters of boron atoms exhibit intriguing size-dependent structures and chemical bonding that are different from bulk boron and may lead to new boron-based nanostructures. We report a combined photoelectron spectroscopic and ab initio study of the 22- and 23-atom boron
Kohei Yazaki et al.
Chemical communications (Cambridge, England), 49(16), 1630-1632 (2013-01-23)
A bowl-shaped organic host was prepared by linking two anthracene-embedded bispyridine ligands with two methylene spacers. The water-soluble host has a hemispherical hydrophobic cavity (∼1 nm in diameter) with two cationic methylenebispyridinyl (Lewis acidic) moieties and shows the selective recognition
Jiang-Fei Xu et al.
Organic letters, 15(24), 6148-6151 (2013-11-19)
Dynamic covalent bonds supplied by reversible anthracene dimerization were combined with pillar[5]arene/imidazole host-guest interactions to construct double-dynamic polymers. Heating such polymers (in solution or as a gel) led to depolymerization by dissociation of either the host-guest complexes alone or the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service