Skip to Content
Merck
All Photos(1)

Documents

566012

Sigma-Aldrich

3-Acetamidophenylboronic acid

≥95%

Synonym(s):

3-Acetamidobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4B(OH)2
CAS Number:
Molecular Weight:
178.98
Beilstein:
3278316
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

mp

135 °C (lit.)

SMILES string

CC(=O)Nc1cccc(c1)B(O)O

InChI

1S/C8H10BNO3/c1-6(11)10-8-4-2-3-7(5-8)9(12)13/h2-5,12-13H,1H3,(H,10,11)

InChI key

IBTSWKLSEOGJGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Suzuki-Miyaura coupling reactions
  • Trifluoromethylation

Reactant involved in the synthesis of a variety of inhibitors including:
  • NR2B subtype of NMDA receptor antagonists for antidepressant activity
  • Biphenylylmethylimidazole derivatives for use as 17,20-lyase inhibitors
  • (Indolyl)-3,5-substituted benzene analogs with antimitotic and antitumor activity
  • Substituted pyrrolidines and tetrahydrofurans as AMPA receptor positive modulators

Other Notes

Contains varying amounts of anhydride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André J van der Vlies et al.
Bioconjugate chemistry, 30(3), 861-870 (2019-01-25)
Curcumin (Cur) has a wide range of bioactivities that show potential for the treatment of cancer as well as chronic diseases associated with inflammation and aging. However, the therapeutic efficacy of Cur has been hampered by its rapid degradation under

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service