Skip to Content
Merck
3-Acetamidophenylboronic acid ≥95%

3-Acetamidophenylboronic acid

3-Acetamidophenylboronic acid ≥95%
Synonym(s):
3-Acetamidobenzeneboronic acid
Linear Formula:
CH3CONHC6H4B(OH)2
CAS Number:
Molecular Weight:
178.98

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing