Skip to Content
Merck
All Photos(1)

Key Documents

547115

Sigma-Aldrich

trans-2-Octen-1-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2OH
CAS Number:
Molecular Weight:
128.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

refractive index

n20/D 1.4460 (lit.)

bp

85-87 °C/10 mmHg (lit.)

density

0.843 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

[H]\C(CO)=C(\[H])CCCCC

InChI

1S/C8H16O/c1-2-3-4-5-6-7-8-9/h6-7,9H,2-5,8H2,1H3/b7-6+

InChI key

AYQPVPFZWIQERS-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

198.0 °F - closed cup

Flash Point(C)

92.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Navideh Sadoughi et al.
Journal of agricultural and food chemistry, 63(11), 2877-2885 (2015-02-24)
An optimized method for the quantitation of volatile compounds responsible for off-aromas, such as earthy odors, found in wine and grapes was developed. The method involved a fast and simple headspace solid-phase microextraction-gas chromatography-mass spectrometry (HS-SPME-GC-MS) for simultaneous determination of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service