Skip to Content
Merck
All Photos(2)

Key Documents

132667

Sigma-Aldrich

trans-2-Hexen-1-ol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCH2OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
1719709
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

96%

form

liquid

refractive index

n20/D 1.438 (lit.)

bp

158-160 °C (lit.)

density

0.849 g/mL at 25 °C (lit.)

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

[H]\C(CO)=C(\[H])CCC

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h4-5,7H,2-3,6H2,1H3/b5-4+

InChI key

ZCHHRLHTBGRGOT-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-2-Hexen-1-ol undergoes allylic epoxidation to yield (2R,3R)-(+)-3-propyloxiranemethanol in high pressure carbon dioxide[1].

Application

trans-2-Hexen-1-ol was used to evaluate the quality of protected designation of virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis[2]. It was also used in encapsulation of vanadium catalysts in inorganic and hybrid matrices using sol-gel method[3].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encapsulation of vanadium complexes in inorganic or hybrid matrices via the sol-gel method: application to the epoxidation of allylic alcohols.
Pellegrino RB and Buffon R.
Journal of the Brazilian Chemical Society, 15(4), 527-531 (2004)
Multicomponent phase equilibrium measurements and modeling for the allylic epoxidation of trans-2-hexen-1-ol to (2R, 3R)-(+)-3-propyloxiranemethanol in high-pressure carbon dioxide.
Stradi BA, et al.
Journal of Supercritical Fluids, 20(1), 1-13 (2001)
Jiaxing Fang et al.
Journal of experimental botany, 71(19), 6084-6091 (2020-06-27)
The symbiosis between the bark beetle (Ips subelongatus) and its fungal symbiont (Endoconidiophora fujiensis) poses a serious threat to larch forests. However, the signaling pathways between these symbiotic partners and their host/non-host trees are not fully understood. Inoculation of the
Benoit Berlioz et al.
Journal of agricultural and food chemistry, 54(26), 10092-10101 (2006-12-21)
Headspace solid-phase microextraction (HS-SPME) -gas chromatography using flame ionization detection and multivariate analysis were applied to the study of the specificity of protected designation of origin (PDO) virgin olive oils produced in a southern French region (Alpes-Maritimes) based on their
Xiaotong Lyu et al.
Food chemistry, 346, 128914-128914 (2021-01-09)
The antioxidants sulfur dioxide (50 ppm) and ascorbic acid (100 ppm) were added to grapes soon after harvest at crushing. The chemical composition and sensory profile of Sauvignon Blanc, Pinot Gris and Chardonnay wines were examined, made from grapes collected at three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service