Skip to Content
Merck
All Photos(1)

Key Documents

546127

Sigma-Aldrich

Coumarin 30

Dye content 99 %

Synonym(s):

3-(2-N-Methylbenzimidazolyl)-7-N,N-diethylaminocoumarin, Coumarin 515

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3O2
CAS Number:
Molecular Weight:
347.41
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 99%

mp

225-229 °C (lit.)

λmax

413 nm

fluorescence

λem 478 nm in ethanol (Lasing peak 510 nm, lasing range 492 - 550 nm (ethylene glycol), pump source Ar (458 nm))

SMILES string

CCN(CC)c1ccc2C=C(C(=O)Oc2c1)c3nc4ccccc4n3C

InChI

1S/C21H21N3O2/c1-4-24(5-2)15-11-10-14-12-16(21(25)26-19(14)13-15)20-22-17-8-6-7-9-18(17)23(20)3/h6-13H,4-5H2,1-3H3

InChI key

KZFUMWVJJNDGAU-UHFFFAOYSA-N

General description

Coumarin 30 (C30) is a laser dye that has a fluorescence yield that tends to unity, which can be potentially used in optical communications and sensors. It is highly stable on the spin coating and shows high quantum efficiency upon low concentration doping.

Application

C30 can form a thin film on p-silicon (p-Si) semiconductor, which can be used to fabricate the aluminum/C30/p-Si based diodes. It can be used as a donor with fluorescein as an acceptor to study the time resolved fluorescence resonance energy transfer (FRET).
Laser dye

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrical and photoelectrical characterization of an organic-inorganic heterojunction based on quinoline yellow dye.
Ugur A, et al.
Materials Science in Semiconductor Processing, 39(8), 569-574 (2015)
Fabrication and electrical properties of an organic-inorganic device based on Coumarin 30 dye.
Tombak A, et al.
Materials Science in Semiconductor Processing, 24(8), 187-192 (2014)
Effect of coumarin on blue light-emitting diodes based on carbazol polymers.
Gautier-Thianche E, et al.
Journal of Applied Physics, 83(8), 4236-4241 (1998)
The nature of photoinduced intermolecular charger transfer in fluorescence resonance energy transfer.
Zong H, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 209(8), 228-233 (2019)
Photophysical Properties of Coumarin-30 Dye in Aprotic and Protic Solvents of Varying Polarities.
Senthilkumar S, et al.
Photochemistry and Photobiology, 80(1), 104-111 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service