Skip to Content
Merck
All Photos(2)

Key Documents

187240

Sigma-Aldrich

9-Anthracenemethanol

97%

Synonym(s):

9-(Hydroxymethyl)anthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O
CAS Number:
Molecular Weight:
208.26
Beilstein:
1873402
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

solubility

chloroform: soluble 20 mg/mL, clear to slightly hazy, light yellow to yellow

functional group

hydroxyl

SMILES string

OCc1c2ccccc2cc3ccccc13

InChI

1S/C15H12O/c16-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)15/h1-9,16H,10H2

InChI key

JCJNNHDZTLRSGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

9-Anthracenemethanol participates in ring-opening polymerization of L-lactide catalyzed by alumoxane[1]. It undergoes proton exchange reaction with potassium tert-butoxide to yield potassium 9-anthracenemethoxide[2].

9-Anthracenemethanol is a derivative of anthracene used in the Diels-Alder reactions. [3]

Application

9-Anthracenemethanol can be used:
  • As a starting material to prepare 9-anthracenylmethyl-1-piperazinecarboxylate, which acts as a reagent in the determination of isocyanates using HPLC.[4]
  • In the Diels-Alder reaction with dimethylacetylene-dicarboxylate to yield lactone derivatives.[5]
  • As an initiator in the ring-opening polymerization of δ-valerolactone to yield poly(δ-valerolactone).[6]
  • As a starting material in the synthesis of polymer-supported anthracene, which acts as a dienophile scavenger in cycloaddition reactions.[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a polymer-supported anthracene and its application as a dienophile scavenger
Lei X and Porco JA
Organic Letters, 6, 795-798 (2004)
J W Flesher et al.
Biochemical and biophysical research communications, 251(1), 239-243 (1998-10-29)
The role of electrophilic hydroxymethyl sulfate esters in the metabolic activation, DNA-damage, mutagenicity, and complete carcinogenicity of polycyclic aromatic hydrocarbons has been elucidated considerably in recent years. The observations are in agreement with a unified hypothesis which predicts that electrophilic
Tamuka Chidanguro et al.
Dalton transactions (Cambridge, England : 2003), 47(26), 8663-8669 (2018-06-23)
We report on the use of visible light as the driving force for the intramolecular dimerization of pendant anthracene groups on a methacrylic polymer to induce the formation of single-chain nanoparticles (SCNPs). Using a 532 nm green laser light source
Yasuhiro Ando et al.
Lipids, 42(10), 955-960 (2007-08-19)
This paper proposes a one-step method for preparation of fluorescent 9-anthrylmethyl esters from triacylglycerols (TAG) ranging in amount from 0.1 to 5 microg. It involves base-catalyzed transesterification using potassium 9-anthracenemethoxide, prepared by proton exchange between 9-anthracenemethanol and potassium tert-butoxide. Transesterification
Yuzhan Li et al.
Scientific reports, 10(1), 20214-20214 (2020-11-21)
The development of multi-stimuli-responsive shape memory polymers has received increasing attention because of its scientific and technological significance. In this work, epoxy elastomers with reversible crosslinks are synthesized by polymerizing an anthracene-functionalized epoxy monomer, a diepoxy comonomer, and a dicarboxylic

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service