Skip to Content
Merck
All Photos(3)

Key Documents

543047

Sigma-Aldrich

Deuterium chloride solution

35 wt. % in D2O, ≥99 atom % D

Synonym(s):

Hydrochloric acid-d solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCl
CAS Number:
Molecular Weight:
37.47
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % D

form

liquid

concentration

35 wt. % in D2O

technique(s)

NMR: suitable

application(s)

pharmaceutical

mass shift

M+1

storage temp.

room temp

SMILES string

Cl[2H]

InChI

1S/ClH/h1H/i/hD

InChI key

VEXZGXHMUGYJMC-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium deuteride is isotopically labeled analogue of Lithium hydride.

Application

Deuterium chloride solution has been used to study:
  • The kinetics of chitosan reacetylation using acetic anhydride and acetic acid.
  • Regioselective terminal group activation of chitosan by thioacetylation.
  • The complex formation between cyclodextrin and captopril in aqueous solution using NMR spectroscopy.

Lithium deuteride can act as semiconductor at 30GPa and room temperature. It is used as the polarized deuteron target material for the measurement of proton and neutron structure

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inclusion complex formation of captopril with $\alpha$-and $\beta$-cyclodextrins in aqueous solution: NMR spectroscopic and molecular dynamic studies.
Ikeda Y, et al.
Journal of Pharmaceutical Sciences, 91(11), 2390-2398 (2002)
Jean Lesne et al.
Nature communications, 11(1), 6140-6140 (2020-12-03)
Hydrogen-Deuterium eXchange coupled to Mass Spectrometry (HDX-MS) is now common practice in structural biology. However, it is most of the time applied to rather small oligomeric complexes. Here, we report on the use of HDX-MS to investigate conformational differences between
Kinetics and efficiency of chitosan reacetylation.
Lavertu M, et al.
Carbohydrate Polymers, 87(2), 1192-1198 (2012)
Regioselective chitosan end-group activation: the triskelion approach
Pickenhahn VD, et al.
Royal Society of Chemistry Advances, 7(30), 18628-18638 (2017)
Andrey N Pravdivtsev et al.
PloS one, 15(10), e0239982-e0239982 (2020-10-02)
Magnetic resonance spectroscopy (MRS) allows the analysis of biochemical processes non-invasively and in vivo. Still, its application in clinical diagnostics is rare. Routine MRS is limited to spatial, chemical and temporal resolutions of cubic centimetres, mM and minutes. In fact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service