Skip to Content
Merck
All Photos(5)

Key Documents

151785

Sigma-Aldrich

Acetic acid-d4

≥99.5 atom % D

Synonym(s):

Acetic-d3 acid-d, Tetradeuteroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CO2D
CAS Number:
Molecular Weight:
64.08
Beilstein:
1748971
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.07 (vs air)

vapor pressure

11.4 mmHg ( 20 °C)

isotopic purity

≥99.5 atom % D

Assay

≥99% (CP)

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %

technique(s)

NMR: suitable
bio NMR: suitable

impurities

≤0.0500% water
water

refractive index

n20/D 1.368 (lit.)

bp

115.5 °C (lit.)

mp

15-16 °C (lit.)

density

1.119 g/mL at 25 °C (lit.)

mass shift

M+4

SMILES string

[2H]OC(=O)C([2H])([2H])[2H]

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1D3/hD

InChI key

QTBSBXVTEAMEQO-GUEYOVJQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetic acid-d4 (Tetradeuteroacetic acid), a deuterated carboxylic acid, is the tetradeuterated form of acetic acid. Its dissociation constant has been evaluated in deuterium oxide (D2O) by an emf method.

Application

Acetic acid-d4 has been used as a deuterated solvent for the dissolution of internal standard in qNMR (Quantitative Nuclear Magnetic Resonance) quantification experiments.
Acetic acid-d4 may also be used as a deuterated solvent in the 1H NMR spectral studies of the following:
  • CB2, a CNBr peptide
  • α - tocopherol succinate hydrophobically modified chitosan(CS-TOS)

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariel Bier et al.
Nutrients, 12(1) (2020-01-17)
High fructose consumption is one of the hallmarks of Western diets and has been found to induce MeS symptoms in parallel to gut microbial dysbiosis. However, the causality between those two is still elusive. Here, we studied whether a significant
a-Tocopherol succinate-modified chitosan as a micellar delivery system for paclitaxel: preparation, characterization and in vitro/in vivo evaluations
Liang N, et al.
International Journal of Pharmaceutics, 423(2), 480-488 (2012)
H Onodera et al.
Natural toxins, 5(4), 146-151 (1997-01-01)
Along with decarbamoylsaxitoxin and decarbamoylgonyautoxin-2 and -3, six new saxitoxin analogues were isolated from the freshwater mat-forming filamentous cyanobacterium Lyngbya wollei collected from Guntersville Reservoir on the Tennessee River in Alabama. Their structures were determined by electrospray ionization mass spectrometry
Ariel Bier et al.
Nutrients, 10(9) (2018-08-26)
Emerging data indicate a correlation between gut microbial composition and cardiovascular disease including hypertension. The host's diet greatly affects microbial composition and metabolite production. Short chain fatty acids (SCFAs) are products of microbial fermentation, which can be utilized by the
Reaction of 13N produced by recoil deuterons in pile-irradiated acetic acid-d4 and malonic acid-d4.
Sensui Y, et al.
J. Radioanal. Nucl. Chem., 118(1), 23-31 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service