Skip to Content
Merck
All Photos(2)

Key Documents

450510

Sigma-Aldrich

Deuterium oxide

99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt

Synonym(s):

Heavy water, Water-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.9 atom % D

Quality Level

form

liquid

contains

0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt

technique(s)

NMR: suitable

bp

101.4 °C (lit.)

mp

3.8 °C (lit.)

SMILES string

[2H]O[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Deuterium oxide (D2O, heavy water) is deuterated water containing 0.05wt.% 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt. It is safe and inexpensive source of deuterium (D) for the D/H exchange reactions. The equilibrium constant for the exchange of deuterium between hydrogen (H) and D2O molecules has been determined. Reports suggest that gelatin gels prepared in D2O show higher rigidity than gels prepared in water. The influence of temperature on the heat capacity of deuterium in crystalline and liquid state was studied calorimetrically and its melting point, enthalpy and entropy of fusion were determined. Its utility as a potential therapeutic agent in treating human pancreatic cancer has been studied.

Application

Deuterium oxide may be used in the following processes:
  • As an NMR solvent.
  • Preparation of β-deuterated alcohols.
  • For the adsorption of neutrons in atomic reactors.
  • Deuterium transfer agent in combination with hexamethyldisilane in the preparation of 1,2-dideuterioalkenes from alkynes.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew J Jezewski et al.
Frontiers in cellular and infection microbiology, 11, 730413-730413 (2021-10-05)
Glycolysis controls cellular energy, redox balance, and biosynthesis. Antiglycolytic therapies are under investigation for treatment of obesity, cancer, aging, autoimmunity, and microbial diseases. Interrupting glycolysis is highly valued as a therapeutic strategy, because glycolytic disruption is generally tolerated in mammals.
Yu-Hsi Lin et al.
Nature metabolism, 2(12), 1413-1426 (2020-11-25)
Inhibiting glycolysis remains an aspirational approach for the treatment of cancer. We have previously identified a subset of cancers harbouring homozygous deletion of the glycolytic enzyme enolase (ENO1) that have exceptional sensitivity to inhibition of its redundant paralogue, ENO2, through
NMR spectroscopy of saccharide-doped PAGAT dosimeters.
Skyt PS, et al.
Journal of Physics. Conference Series, 573 (2015)
The Exchange Reaction of Hydrogen and Deuterium Oxide.
Crist RH and Dalin GA
J. Chem. Phys. , 2(8), 548-548 (1934)
Thermodynamic properties of deuterium oxide in the temperature range from 6 to 350K.
Smirnova NN, et al.
The Journal of Chemical Thermodynamics, 38(7), 879-883 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service