Skip to Content
Merck
All Photos(2)

Key Documents

534498

Sigma-Aldrich

4-Piperidinopiperidine

97%

Synonym(s):

1,4′-Bipiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2
CAS Number:
Molecular Weight:
168.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

64-66 °C (lit.)

storage temp.

−20°C

SMILES string

C1CCN(CC1)C2CCNCC2

InChI

1S/C10H20N2/c1-2-8-12(9-3-1)10-4-6-11-7-5-10/h10-11H,1-9H2

InChI key

QDVBKXJMLILLLB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Medicinal chemistry reagent.
Reactant for synthesis of:
Arylthiadiazole H3 antagonists
Water-soluble N-mustards as anticancer agents
Antitubercular drugs
Vasopressin1b receptor antagonists
MDR modulators
Selective Norepinephrine transporter inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuqiang Wang et al.
Bioorganic & medicinal chemistry, 13(19), 5592-5599 (2005-08-09)
We have synthesized a conjugate of cis-4,7,10,13,16,19-docosahexenoic acid (DHA) and 10-hydroxycamptothecin (HCPT), DHA-HCPT. The antitumor activity of DHA-HCPT was evaluated in vitro against L1210 leukemia cells and in experimental animal tumor models including L1210 leukemia, Lewis lung carcinoma, and colon
Synlett, 8, 1192-1194 (2003)
Janice L Hyatt et al.
Chemico-biological interactions, 157-158, 247-252 (2005-11-01)
CPT-11 (irinotecan, 7-ethyl-10-[4-(1-piperidino)-1-piperidino]carbonyloxycamptothecin) is an anticancer prodrug that has been approved for the treatment of colon cancer. It is a member of the camptothecin class of drugs and activation to the active metabolite SN-38, is mediated by carboxylesterases (CE). SN-38
Hidetsura Cho et al.
Journal of medicinal chemistry, 47(1), 101-109 (2003-12-30)
A variety of novel heterocyclic compounds having thienoazepine, pyrroloazepine, furoazepine, and thienodiazepine skeletons were synthesized, most of which exhibited potent antagonism of [(3)H]-AVP specific binding in assays using rat liver (V1), rat kidney (V2), human platelet plasma membranes, and recombinant
A W McConnaughie et al.
Journal of medicinal chemistry, 37(8), 1063-1069 (1994-04-15)
As as initial step in the design of structure-specific RNA-interactive molecules as potential antiviral agents, we have focused on the synthesis of molecules that exhibit strong and preferential binding to duplex RNA. A series of polycationic ligands have been synthesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service