497312
4-Piperidinemethanol
97%
Synonym(s):
4-(Hydroxymethyl)piperidine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
bp
118-120 °C/10 mmHg (lit.)
mp
55-59 °C (lit.)
functional group
hydroxyl
SMILES string
OCC1CCNCC1
InChI
1S/C6H13NO/c8-5-6-1-3-7-4-2-6/h6-8H,1-5H2
InChI key
XBXHCBLBYQEYTI-UHFFFAOYSA-N
Related Categories
General description
4-Piperidinemethanol is a cyclic secondary amine. Its standard molar enthalpies of combustion, sublimation and formation have been determined.
Application
4-Piperidinemethanol may be used in the preparation of:
- N-tert-butoxycarbonyl-4-hydroxymethyl piperidine
- desferrioxamine B (DFO) containing third generation triazine dendrimer
- ethyl 3-(4-(hydroxymethyl)piperidin-1-yl)propanoate (EHMPP)
- 4-(hydroxymethyl)piperidine-1-carbodithioic acid (HL)
- 1-[[(1E)-2-(4-chlorophenyl)ethenyl]sulfonyl]-4-piperidinemethanol
Substrate used in solid-phase organic synthesis of a secondary amine.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jongdoo Lim et al.
Bioorganic & medicinal chemistry, 18(15), 5749-5753 (2010-07-10)
The synthesis of a third generation triazine dendrimer, 1, containing multiple, iron-sequestering desferrioxamine B (DFO) groups is described. Benzoylation of the hydroxamic acid groups of DFO and formation of a reactive dichlorotriazine provide the intermediate for reaction with the second
Enzymatic synthesis, thermal and crystalline properties of a poly (β-amino ester) and poly (lactone-co-β-amino ester) copolymers.
Martino L, et al.
Polymer, 53(9), 1839-1848 (2012)
Synthesis and evaluation of 1-arylsulfonyl-3-piperazinone derivatives as factor Xa inhibitor.
Nishida H, et al.
Chemical & Pharmaceutical Bulletin, 49(10), 1237-1244 (2001)
Christian A Olsen et al.
Organic letters, 6(12), 1935-1938 (2004-06-05)
[reaction: see text] An expedient solid-phase synthetic approach to secondary and tertiary amines was developed. The protocol employs conversion of resin-bound amino alcohols to the corresponding iodides, followed by iodide displacement with primary or secondary amines or with unprotected amino
Francis Giraud et al.
Bioorganic & medicinal chemistry letters, 19(2), 301-304 (2008-12-19)
Continuous efforts on the synthesis and structure-activity relationships (SARs) studies of modified 1-benzylamino-2-phenyl-3-(1H-1,2,4-triazol-1-yl)propan-2-ols as antifungal agents, allowed identification of new 1-[(pyridinyl- and piperidinylmethyl)amino] derivatives with MIC(80) values ranging from 1410.0 to 23.0ngmL(-1) on Candidaalbicans. These results confirmed both the importance
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service