All Photos(2)
About This Item
Linear Formula:
FC6H4C≡CH
CAS Number:
Molecular Weight:
120.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.5170 (lit.)
bp
138 °C (lit.)
density
1.039 g/mL at 25 °C (lit.)
functional group
fluoro
SMILES string
Fc1cccc(c1)C#C
InChI
1S/C8H5F/c1-2-7-4-3-5-8(9)6-7/h1,3-6H
InChI key
PTRUTZFCVFUTMW-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
90.0 °F - closed cup
Flash Point(C)
32.2 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Gold film-catalysed benzannulation by microwave-assisted, continuous flow organic synthesis (MACOS).
Gjergji Shore et al.
Beilstein journal of organic chemistry, 5, 35-35 (2009-09-25)
Methodology has been developed for laying down a thin gold-on-silver film on the inner surface of glass capillaries for the purpose of catalysing benzannulation reactions. The cycloaddition precursors are flowed through these capillaries while the metal film is being heated
Lei Zhou et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(43), 11668-11674 (2009-09-25)
A simple and efficient method for the synthesis of gamma,delta-alkynyl-beta-amino acid derivatives by a copper-catalyzed three-component amine-alkyne-alkyne addition reaction was developed. Various gamma,delta-alkynyl-beta-amino acid derivatives were synthesized in moderate to good yields in one step. With chiral prolinol derivatives employed
Ya Zhou et al.
Probe Reports from the NIH Molecular Libraries Program, 2011 Oct 31 (Updated 2013 Mar 7) (2013-06-14)
A series of acetylenic biaryl mGlu5 positive allosteric modulators (PAMs) have been optimized as pure potentiators in low receptor expressing mGlu5 cell lines. ML254 was identified and shown to competitively interact with the MPEP allosteric binding site. Preliminary data from
Jason Manka et al.
Probe Reports from the NIH Molecular Libraries Program, 2011 Dec 16 (Updated 2013 Mar 7) (2013-06-14)
Allosteric modulators for G-protein-coupled receptors (GPCRs) provide numerous advantages over orthosteric ligands, including greater sub-type selectivity, reduced receptor desensitization, saturability of effect, and potential for enhanced therapeutic index. Positive allosteric modulators (PAMs) of the group I metabotropic glutamate receptor mGlu
(NHC) Copper (I)-Catalyzed [3+ 2] Cycloaddition of Azides and Mono-or Disubstituted Alkynes.
Diez-Gonzalez S, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 12(29), 7558-7564 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service