Skip to Content
Merck
All Photos(1)

Documents

465305

Sigma-Aldrich

1-Chloro-2-ethynylbenzene

98%

Synonym(s):

(2-Chlorophenyl)acetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4C≡CH
CAS Number:
Molecular Weight:
136.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.5710 (lit.)

bp

82 °C/12 mmHg (lit.)

density

1.125 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1ccccc1C#C

InChI

1S/C8H5Cl/c1-2-7-5-3-4-6-8(7)9/h1,3-6H

InChI key

DGLHLIWXYSGYBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Chloro-2-ethynylbenzene is a chlorinated benzene derivative.

Application

1-Chloro-2-ethynylbenzene may be used in the synthesis of 4-(2-chlorophenylethynyl)-3-methyl-5-phenylisoxazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 3, 4, 5-Trisubstituted Isoxazoles through Gold-Catalyzed Cascade Cyclization- Oxidative Alkynylation and Cyclization-Fluorination of 2-Alkynone O-Methyloximes.
Song D-H and Ryu J-S.
Bull. Korean Chem. Soc., 35(9), 2635-2644 (2014)
Comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry and simultaneous electron capture detection/nitrogen phosphorous detection for incense analysis.
Tran TC and Marriott PJ.
Atmospheric Environment, 42(32), 7360-7372 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service