Skip to Content
Merck
All Photos(3)

Key Documents

471402

Sigma-Aldrich

1-Hexanol

anhydrous, ≥99%

Synonym(s):

Hexyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
969167
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
156-157 °C (lit.)
vapor pressure:
1 mmHg ( 25.6 °C)

grade

anhydrous

Quality Level

vapor density

4.5 (vs air)

vapor pressure

1 mmHg ( 25.6 °C)

Assay

≥99%

form

liquid

autoignition temp.

559 °F

expl. lim.

0.34-6.3 %

impurities

<0.005% water

evapn. residue

<0.0005%

refractive index

n20/D 1.418 (lit.)

bp

156-157 °C (lit.)

mp

−52 °C (lit.)

density

0.814 g/mL at 25 °C (lit.)

SMILES string

CCCCCCO

InChI

1S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3

InChI key

ZSIAUFGUXNUGDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexanol is a linear primary alcohol. It is formed as an intermediate during the catalytic transformation of cellulose. The ability of 1,1,3,3-tetramethylguanidine (TMG) in 1-hexanol solvent system to capture carbon dioxide has been assessed. The solubility of light fullerenes in 1-hexanol as a function of temperature and pressure was studied.
1-Hexanol is produced from coconut oil and palm oils. It is used in the production of antiseptics, fragrances and perfumes. 1-Hexanol is also used as a solvent in the production of plasticizers.

Application

1-Hexanol has been used as an odorant to study olfactory responses and to thin the dielectric layer of poly(4-vinylphenol) (PVP).

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct catalytic conversion of cellulose to liquid straight-chain alkanes.
de Beeck BO, et al.
Energy & Environmental Science, 8(1), 230-240 (2015)
Shalene Xue Lin Goh et al.
Analytica chimica acta, 1035, 77-86 (2018-09-19)
A novel fully automated liquid-phase microextraction (LPME) procedure making use of a conical polypropylene membrane bag to hold the solvent, coupled with ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) with positive electrospray ionisation was developed to determine glucocorticoids (including cortisol
A 4-dimensional representation of antennal lobe output based on an ensemble of characterized projection neurons
Staudacher EM, et al.
Journal of Neuroscience Methods, 180(2), 208-223 (2009)
Patty's Toxicology (2012)
Kinetics of carbon dioxide binding by 1,1,3,3-tetramethylguanidine in 1-hexanol.
Ozturk MC, et al.
International Journal of Greenhouse Gas Control, 26, 76-82 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service