Skip to Content
Merck
All Photos(1)

Key Documents

514004

Sigma-Aldrich

1-Azido-1-deoxy-β-D-glucopyranoside

Synonym(s):

Β-D-Glycopyranosyl azide, 1-Azido-1-deoxy-beta-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3O5
CAS Number:
Molecular Weight:
205.17
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: click chemistry

mp

60-65 °C (lit.)

SMILES string

OC[C@H]1O[C@@H](N=[N+]=[N-])[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H11N3O5/c7-9-8-6-5(13)4(12)3(11)2(1-10)14-6/h2-6,10-13H,1H2/t2-,3-,4+,5-,6-/m1/s1

InChI key

KSRDTSABQYNYMP-VFUOTHLCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Azido-1-deoxy-β-D-glucopyranoside can be used as an organic building block to prepare:
  • 1,2,3-triazole-containing (1-azido-1-deoxy-β-D-glucopyranoside) complex by reacting with propagylglycine and sodium ascorbate via copper-catalyzed click chemistry approach.
  • D-glucose-mercaptoacetyl triglycine (MAG3) derivative applicable as a potent tumor diagnosis agent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André Luís Branco de Barros et al.
Bioorganic & medicinal chemistry letters, 19(9), 2497-2499 (2009-04-01)
A d-glucose-MAG(3) derivative was successfully synthesized and radiolabeled in high labeling yield. Biodistribution studies in Ehrlich tumor-bearing mice were performed. This compound showed high accumulation in tumor tissue with high tumor-to-muscle ratio and moderate tumor-to-blood ratio. Thus, d-glucose-MAG(3) is a
IrfanUllah Khan et al.
Pakistan journal of pharmaceutical sciences, 29(1), 213-219 (2016-02-02)
The 1,2,3-triazole-containing (1-azido-1-deoxy-β-D-glucopyranoside) complex was synthesized using click chemistry approach and evaluated its potential as a tumor-seeking agent. In the present study, (99m)Tc-tricarbonyl labeled (1-azido-1-deoxy-β-D-glucopyranoside) radiotracer [(99m)Tc(CO)(3)-BM], (where BM stands for biomolecule, e.g., (1-azido-1-deoxy-β-D- glucopyranoside)) was synthesized via click chemistry
S K Sinha et al.
Carbohydrate research, 81(2), 239-247 (1980-03-15)
Syntheses are reported of 4-deoxy-D-xylo-hexose and 4-azido-4-deoxy-D-glucose as potential inhibitors for lactose synthase [uridine 5'-(alpha-D-galactopyranosyl pyrophosphate):D-glucose 4-beta-D-galactopyranosyltransferase, EC 2.4.1.22]. These syntheses involved SN2 displacement of the 4-methylsulfonyloxy group of methyl 2,3,6-tri-O-benzoyl-4-O-methylsulfonyl-alpha-D-galactopyranoside by iodide and azide ions. In both cases, inversion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service