Skip to Content
Merck
All Photos(1)

Key Documents

494372

Sigma-Aldrich

Ytterbium(II) iodide

powder, ≥99.9% trace metals basis

Synonym(s):

Ytterbium diiodide, Ytterbium(II) iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YbI2
CAS Number:
Molecular Weight:
426.85
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.9% trace metals basis
form:
powder

Quality Level

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: ytterbium

impurities

≤1000.0 ppm Trace Rare Earth Analysis

mp

~999 °C (lit.)

SMILES string

I[Yb]I

InChI

1S/2HI.Yb/h2*1H;/q;;+2/p-2

InChI key

SJLISRWUWZVXNZ-UHFFFAOYSA-L

Application


  • Divalent Ytterbium Iodide Supported by beta-Diketiminato Based Tridentate Ligand: Synthesis, Structure and Small Molecule Activation: This study explores the synthesis of an ytterbium(II) iodide supported by a beta-diketiminato ligand, highlighting its structure and potential in small molecule activation (X Liu, et al., 2020).

  • Chemical Engineering of Quasicrystal Approximants in Lanthanide-Based Coordination Solids: This paper discusses the use of ytterbium(II) as a building block in quasicrystal approximants with coordination solids, showing how iodide ligands contribute to the stability and assembly of these structures (L Voigt, et al., 2020).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service