Skip to Content
Merck
All Photos(2)

Key Documents

347116

Sigma-Aldrich

Samarium(II) iodide solution

0.1 M in THF, contains samarium chips as stabilizer

Synonym(s):

Samarium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SmI2
CAS Number:
Molecular Weight:
404.17
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

contains

samarium chips as stabilizer

reaction suitability

core: samarium
reagent type: catalyst
reaction type: Reductions

reagent type: reductant

concentration

0.1 M in THF

density

0.922 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

I[Sm]I

InChI

1S/2HI.Sm/h2*1H;/q;;+2/p-2

InChI key

UAWABSHMGXMCRK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Samarium(II) iodide solution (SmI2) can be used as a reagent in the synthesis of:
  • Benzannulated pyrrolizidines and indolizidines by SmI2-induced cyclizations of indole derivatives.
  • Chiral 4-substituted 2-oxazolidinones and 5,5-disubstituted oxazolidinones through asymmetric Reformatsky type reaction.
  • γ-Aminoalkyl substituted γ-butyrolactones via ketyl-alkene coupling reaction.

It can also be used in:
  • Reduction of conjugated double and triple bonds into alkenes using SmI2/H2O/amine mixtures.
  • Conversion of β-hydroxyketones into 1,3-diols by SmI2/H2O/Et3N.
  • Selective reduction of 6-membered lactones to the corresponding diols/triols using SmI2-H2O reagent system.

SmI2 is an effective single-electron reducing agent for the promotion of ketone-olefin, ketyl aryl cyclizations, and pinacol coupling reactions under mild conditions. Often both intramolecular and intermolecular couplings proceed in a highly stereoselective fashion. It is also used in the synthesis of new heteroleptic samarium aryloxide/cyclopentadienide complexes.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3 - Water-react 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup - Solvent

Flash Point(C)

-19 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-Step Highly Diastereoselective Synthesis of γ-Aminoalkyl-Substituted γ-Butyrolactones by an Asymmetric Samarium-Mediated Ketyl-Alkene Coupling Reaction
Fukuzawa S-I, et al.
The Journal of Organic Chemistry, 68(5), 2042-2044 (2003)
Tetrahedron, 59, 10351-10351 (2003)
Reduction of β-Hydroxyketones by SmI2/H2O/Et3N
Davis TA, et al.
Organic Letters, 7(1), 119-122 (2005)
Samarium(II) Iodide Promoted Intermolecular Ketone-Olefin Couplings Chelation-Controlled by .alpha.-Hydroxyl Groups
Motoi Kawatsura FM and Haruhisa Shirahama
The Journal of Organic Chemistry, 59, 6900-6900 (1994)
Zhaomin Hou et al.
Inorganic chemistry, 35(25), 7190-7195 (1996-12-04)
Synthesis of a new class of heteroleptic samarium aryloxide complexes has been achieved by the use of homoleptic samarium(II) bis(aryloxide) Sm(OAr)(2)(THF)(3) (1, Ar = C(6)H(2)Bu(t)(2)-2,6-Me-4) as a starting material, which is easily obtained by reaction of Sm(N(SiMe(3))(2))(2)(THF)(2) with 2 equiv

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service