474770
Europium(II) iodide
anhydrous, powder, 99.9% trace metals basis
Synonym(s):
Europium diiodide, Europium iodide
About This Item
Recommended Products
grade
anhydrous
Assay
99.9% trace metals basis
form
powder
reaction suitability
reagent type: catalyst
core: europium
impurities
≤1500.0 ppm Trace Rare Earth Analysis
SMILES string
I[Eu]I
InChI
1S/Eu.2HI/h;2*1H/q+2;;/p-2
InChI key
DPYXWFUVSMSNNV-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
- Reactions of strontium, lanthanum, and europium iodides. Direct preparation of phosphorus triiodide from phosphates: Explores reactions and preparation methods involving europium iodides (Haschke, 1976).
accessory
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service