Skip to Content
Merck
All Photos(3)

Key Documents

479411

Sigma-Aldrich

4,6-Dimethyldibenzothiophene

97%

Synonym(s):

4,6-DMDBT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12S
CAS Number:
Molecular Weight:
212.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

153-157 °C (lit.)

SMILES string

Cc1cccc2c3cccc(C)c3sc12

InChI

1S/C14H12S/c1-9-5-3-7-11-12-8-4-6-10(2)14(12)15-13(9)11/h3-8H,1-2H3

InChI key

MYAQZIAVOLKEGW-UHFFFAOYSA-N

General description

4,6-Dimethyldibenzothiophene (4,6-DMDBT), a high refractory sulfur compound, is commonly found in diesel fuel. Its synthesis has been reported. The hydrodesulfurization of 4,6-DMDBT using bulk nickel alloy, bulk tungsten phosphide (WP), NiMo sulfide supported on active carbons and molecularly imprinted polymers have been reported. The dielectric behavior of 4,6-DMDBT under different microwave frequencies and temperature has been investigated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hussein N Nassar et al.
Environmental science and pollution research international, 28(7), 8102-8116 (2020-10-14)
One of the main precursors of air pollution and acid rains is the presence of the recalcitrant thiophenic compounds, for example dibenzothiophene (DBT) and its derivatives in transportation fuels. In an attempt to achieve the worldwide regulations of ultra-low sulfur
Development of a novel molecularly imprinted polymer for the retention of 4,6-dimethyldibenzothiophene.
Tom LA, et al.
Microchimica Acta, 176(3-4), 375-380 (2012)
An efficient synthesis of pure 4,6-dimethyldibenzothiophene.
Meille V, et al.
Tetrahedron, 52(11), 3953-3960 (1996)
Hydrodesulfurization kinetics and mechanism of 4,6-dimethyldibenzothiophene over NiMo catalyst supported on carbon.
Sakanishi K, et al.
J. Mol. Catal. A: Chem., 155(1), 101-109 (2000)
Meiqin Zheng et al.
Journal of hazardous materials, 362, 424-435 (2018-09-28)
In this work, the adsorption desulfurization performance and adsorption diffusion study of B2O3 modified Ag-CeOx/TiO2-SiO2 adsorbent were investigated. The adsorption desulfurization performance was studied by batch and fixed bed tests. The homogeneous surface diffusion model (HSDM) was employed to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service