Skip to Content
Merck
All Photos(2)

Documents

357790

Sigma-Aldrich

Thianaphthene

98%

Synonym(s):

Benzo[b]thiophene, Benzothiophene, Thionaphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6S
CAS Number:
Molecular Weight:
134.20
Beilstein:
80580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

221-222 °C (lit.)

mp

30-33 °C (lit.)

density

1.149 g/mL at 25 °C (lit.)

SMILES string

c1ccc2sccc2c1

InChI

1S/C8H6S/c1-2-4-8-7(3-1)5-6-9-8/h1-6H

InChI key

FCEHBMOGCRZNNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of ethyl diazoacetate with thianaphthene has been reported.

Application

Thianaphthene may be used in the preparation of 2-thianapthenylphenyllithium, via metalation by n-butyllithium.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some 2-Substituted Thianaphthenes Derived from 2-Thianaphthenyllithium1.
Shirley DA and Cameron MD.
Journal of the American Chemical Society, 74(3), 664-665 (1952)
Reactions of ethyl diazoacetate with thianaphthene, indoles, and benzofuran.
Wenkert E, et al.
The Journal of Organic Chemistry, 42(24), 3945-3949 (1977)
Haiying She et al.
Journal of colloid and interface science, 518, 149-155 (2018-02-18)
An adsorbent with multiple interaction sites for the adsorption of nitrogen-containing compounds (NCCs) has been realized in a silver ion functionalized Cr3+ based metal-organic framework (Cr)-MIL-101-SO3Ag. The adsorptive denitrogenation performance of (Cr)-MIL-101-SO3Ag was evaluated in a batch adsorption system in
Eva Tyteca et al.
Journal of chromatography. A, 1358, 145-154 (2014-07-21)
Linear gradient programs are very frequently used in reversed phase liquid chromatography to enhance the selectivity compared to isocratic separations. Multi-linear gradient programs on the other hand are only scarcely used, despite their intrinsically larger separation power. Because the gradient-conformity
Argam Akopyan et al.
Molecules (Basel, Switzerland), 25(3) (2020-01-30)
Polyoxometalate-based ionic liquid hybrid materials with a pyridinium cation, containing Brönsted acid sites, were synthesized and used as catalysts for the oxidation of model and real diesel fuels. Keggin-type polyoxometalates with the formulae [PMo12O40]3-, [PVMo11O40]4-, [PV2Mo10O40]4-, [PW12O40]3- were used as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service