Skip to Content
Merck
All Photos(1)

Documents

471690

Sigma-Aldrich

1,3,2-Dioxathiolane 2,2-dioxide

98%

Synonym(s):

1,2-Ethylene sulfate, 1,3,2λ6-Dioxathiolane-2,2-dione, 1,3,2-Dioxathiolan-2,2-dioxide, Ethylene sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O4S
CAS Number:
Molecular Weight:
124.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

95-97 °C (lit.)

storage temp.

2-8°C

SMILES string

O=S1(=O)OCCO1

InChI

1S/C2H4O4S/c3-7(4)5-1-2-6-7/h1-2H2

InChI key

ZPFAVCIQZKRBGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3,2-Dioxathiolane 2,2-dioxide, also known as Ethylene sulfate, is a cyclic sulfate that is commonly used as an alkylating agents and intermediate in the synthesis of disulfates and sulfonate surfactants.

Application

1,3,2-Dioxathiolane 2,2-dioxide may be employed in the preparation of imidazolidinium salts.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and kinetic study of 1, 3, 2-dioxathiolane 2, 2-dioxide in microreactors
Ting W, et al.
Reaction Chemistry & Engineering (2024)
1, 3, 2-Dioxathiolane 2, 2-Dioxide
Nicholas L J, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis (2001)
Novel, one-pot procedure for the synthesis of 2-arylethanol derivatives
Torsten S, et al.
Synthesis, 2008, 1793-1797 (2008)
Eric Schönemann et al.
Polymers, 11(6) (2019-06-12)
Films of zwitterionic polymers are increasingly explored for conferring fouling resistance to materials. Yet, the structural diversity of polyzwitterions is rather limited so far, and clear structure-property relationships are missing. Therefore, we synthesized a series of new polyzwitterions combining ammonium
Synthesis of hydroxy sulfonate surfactants
Oystein R, et al.
Molecules (Basel), 10, 1169-1178 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service