Skip to Content
Merck
All Photos(2)

Documents

774243

Sigma-Aldrich

1,3-Propylene sulfite

greener alternative

99%

Synonym(s):

2-Oxo-1,3,2-dioxathiane, PS, TMS, Trimethylene sulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3S
CAS Number:
Molecular Weight:
122.14
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.453

bp

73 °C (lit.)

mp

-25 °C (lit.)

density

1.347 g/mL at 25 °C

application(s)

battery manufacturing

greener alternative category

SMILES string

O=S1OCCCO1

InChI

1S/C3H6O3S/c4-7-5-2-1-3-6-7/h1-3H2

InChI key

LOURZMYQPMDBSR-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Trimethylene sulfite is used as high temperature additive for electrolytes in Lithium ion batteries. It improves the decomposition resistance of the electrolyte.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.0 °F

Flash Point(C)

83.9 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahra Rezaei et al.
Bioorganic chemistry, 90, 103055-103055 (2019-06-21)
Structure activity correlation revealed that the quinoxaline ring is a satisfactory backbone for anticancer activity and a specific functional group at position 1 and 2 can improve the activity. In this basis, besides quinoxaline, imidazoles as potential anticancer agents were
Colin Hawco et al.
Frontiers in human neuroscience, 11, 404-404 (2017-09-01)
There has been a distinct shift in neuroimaging from localization of function into a more network based approach focused on connectivity. While fMRI has proven very fruitful for this, the hemodynamic signal is inherently slow which limits the temporal resolution
Stephen P Bailey et al.
International journal of sports physiology and performance, 14(7), 927-933 (2019-01-25)
The purpose of this investigation is to determine the effects of different forms of a CHO MR on quadriceps muscle performance and corticospinal motor excitability. Ten subjects (5 females, 5 males; 25±1 years; 1.71±0.03 m 73±5 kg) completed 4 trials.
Taek Lee et al.
Nanomaterials (Basel, Switzerland), 9(7) (2019-08-03)
In the present study, we fabricated a dual-mode cardiac troponin I (cTnI) biosensor comprised of multi-functional DNA (MF-DNA) on Au nanocrystal (AuNC) using an electrochemical method (EC) and a localized surface plasmon resonance (LSPR) method. To construct a cTnI bioprobe
Kang He et al.
Natural product research, 33(20), 2982-2987 (2018-11-30)
A new xanthone glycoside, 3,5,7,8-tetramethoxyxanthone-1-O-β-D-glucopyranoside (1), along with five known compounds, mangiferin (2), kaempferol (3), quercetin (4), chlorogenic acid (5) and diploptene (6), was isolated from the whole plants of Pyrrosia sheareri (Bak.) Ching. The structure of compound 1 was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service