Skip to Content
Merck
All Photos(1)

Documents

291250

Sigma-Aldrich

1,3-Propanesultone

≥99%

Synonym(s):

3-Hydroxypropanesulfonic acid γ-sultone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3S
CAS Number:
Molecular Weight:
122.14
Beilstein:
109782
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

bp

180 °C/30 mmHg (lit.)

mp

30-33 °C (lit.)

density

1.392 g/mL at 25 °C (lit.)

SMILES string

O=S1(OCCC1)=O

InChI

1S/C3H6O3S/c4-7(5)3-1-2-6-7/h1-3H2

InChI key

FSSPGSAQUIYDCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Propanesultone has been used in:
  • preparation of poly[2-ethynyl-N-(propylsulfonate)pyridinium betaine], a new ionic conjugated polymer
  • preparation of novel poly(4-vinylpyridine) supported acidic ionic liquid catalyst
  • preparation of poly((2-(dimethylamino)ethyl methacrylate)-co-3-dimethyl(methacryloyloxyethyl)ammonium propanesulfonate)-coated mesoporous silica nanoparticles
  • sulfonation of surface of self-assembled nanoporous silica colloidal crystalline films comprised of 184nm-diameter silica spheres

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

340.7 °F

Flash Point(C)

171.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kang-Ting Huang et al.
Biomacromolecules, 20(9), 3524-3534 (2019-08-06)
This work presents a salt-responsive interpenetrating network (IPN) hydrogel with effective antimicrobial properties and surface regeneration. The hydrogels were engineered using the double network strategy to form loosely cross-linked zwitterionic poly(sulfobetaine vinylimidazole) (pSBVI) networks into the highly cross-linked cationic poly((trimethylamino)ethyl
Junke Wang et al.
Ultrasonics sonochemistry, 21(1), 29-34 (2013-06-12)
A novel poly(4-vinylpyridine) supported acidic ionic liquid catalyst was synthesized by the reaction of 4-vinylpyridine with 1,3-propanesultone, followed by the polymerization and the addition of the heteropolyacid. Due to the combination of polymer features and ionic liquid, it acted as
Stephen D Dertinger et al.
Environmental and molecular mutagenesis, 52(9), 748-755 (2011-11-05)
An international collaborative trial was established to systematically investigate the merits and limitations of a rat in vivo Pig-a gene mutation assay. The product of this gene is essential for anchoring CD59 to the plasma membrane, and mutations in this
Hermann M Bolt et al.
Toxicology letters, 151(1), 251-254 (2004-06-05)
1,3-Propane sultone is directly alkylating, genotoxic and carcinogenic. In rats, it induces local and systemic tumours at multiple target sites. Preponderant systemic tumours occur at the central nervous system, especially gliomas. Other localisations include the mammary gland, the intestine, the
D K Torous et al.
Mutation research, 465(1-2), 91-99 (2000-03-10)
Micronuclei (MN) are routinely enumerated in mouse peripheral blood to index genotoxicity. Recent data from the Collaborative Study Group for the Micronucleus Test (CSGMT) [CSGMT (The Collaborative Study Group for the Micronucleus Test), Evaluation of the rat micronucleus test with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service