Skip to Content
Merck
All Photos(2)

Documents

469130

Sigma-Aldrich

Copper(II) sulfate pentahydrate

99.995% trace metals basis

Synonym(s):

Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

7.3 mmHg ( 25 °C)

Assay

99.995% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: copper

impurities

≤0.001% N compounds
≤0.005% insolubles
<50 ppm total metallic impurities

mp

110 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ca: ≤0.005%
Fe: ≤0.003%
K: ≤0.01%
Na: ≤0.02%
Ni: ≤0.005%

application(s)

battery manufacturing

SMILES string

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Copper (II) sulfate pentahydrate salt may be used for the fabrication of copper nanoparticles by chemical reduction. The pentahydrate salt of copper may be used as a catalyst for the conversion of aromatic aldehydes to primary amides via aldoximes. Reduced graphene oxide-supported copper nanoparticles (rGO/Cu NPs) may be prepared by copper (II) sulfate pentahydrate and graphitic precursors. An aqueous electrolytic bath containing CuSO4.5H2O as one of the constituents was used for the preparation of Cu2ZnSnS4 (CZTS) thin film solar cell. Ferric chloride hexahydrate (FeCl3.6H2O) and copper(II) sulfate pentahydrate (CuSO4.5H2O) may be used to fabricate Fe-Cu binary oxide sorbents for arsenic removal applications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient copper (II)-catalyzed direct access to primary amides from aldehydes under neat conditions.
Ganguly NC, et al.
Tetrahedron Letters, 53(11), 1413-1416 (2012)
Synthesis and optical properties of copper nanoparticles prepared by a chemical reduction method.
Dang TMD, et al.
Nanoscience and Nanotechnology, 2(1), 015009-015009 (2011)
Cu2 ZnSnS 4 thin films and nanowires prepared by different single-step electrodeposition method in quaternary electrolyte.
Jeon M, et al.
Materials Letters, 65(15), 2364-2367 (2011)
Nanostructured iron (III)-copper (II) binary oxide: a novel adsorbent for enhanced arsenic removal from aqueous solutions.
Zhang G, et al.
Water Research, 47(12), 4022-4031 (2013)
Synthesis and characterization of copper nanoparticles supported on reduced graphene oxide as a highly active and recyclable catalyst for the synthesis of formamides and primary amines
Fakhri P, et al.
J. Mol. Catal. A: Chem., 383?384, 17-22 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service