Skip to Content
Merck
All Photos(1)

Key Documents

460931

Sigma-Aldrich

Neopentyl chloroformate

97%

Synonym(s):

2,2-Dimethylpropyl chloroformate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2OCOCl
CAS Number:
Molecular Weight:
150.60
Beilstein:
506655
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

−0.65 psi ( 20 °C)

Quality Level

Assay

97%

refractive index

n20/D 1.410 (lit.)

bp

55 °C/36 mmHg (lit.)

density

1.003 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)COC(Cl)=O

InChI

1S/C6H11ClO2/c1-6(2,3)4-9-5(7)8/h4H2,1-3H3

InChI key

JUUBFHLPTCPVBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neopentyl chloroformate, a branched primary alkyl chloroformate, is a useful protecting agent during peptide synthesis.

Application

Neopentyl chloroformate was used in the preparation of mixture of 11,12-dimethyltetradecanoic acid and 11-ethyl-12-methyltridecanoic acid.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malcolm J D'Souza et al.
International journal of molecular sciences, 12(2), 1161-1174 (2011-05-05)
The specific rates of solvolysis of neopentyl chloroformate (1) have been determined in 21 pure and binary solvents at 45.0 °C. In most solvents the values are essentially identical to those for ethyl and n-propyl chloroformates. However, in aqueous-1,1,1,3,3,3-hexafluoro-2-propanol mixtures
Ursula Biermann et al.
Journal of the American Chemical Society, 126(33), 10319-10330 (2004-08-19)
A general method for the hydro-alkyl addition to the nonactivated C=C double bond of alkenes using alkyl chloroformates (primary, secondary), 12, and di-tert-butylpyrocarbonate, 52, mediated by ethylaluminum sesquichloride (Et(3)Al(2)Cl(3)) has been developed. Reaction of 12 and 52, respectively, with Et(3)Al(2)Cl(3)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service