Skip to Content
Merck
All Photos(1)

Key Documents

451304

Sigma-Aldrich

Terbium(III) chloride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Terbium chloride, Terbium trichloride, Terbium(III) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TbCl3
CAS Number:
Molecular Weight:
265.28
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: terbium

impurities

≤150.0 ppm Trace Rare Earth Analysis

mp

588 °C (lit.)

density

4.35 g/mL at 25 °C (lit.)

SMILES string

Cl[Tb](Cl)Cl

InChI

1S/3ClH.Tb/h3*1H;/q;;;+3/p-3

InChI key

GFISHBQNVWAVFU-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Pineda et al.
Biochimica et biophysica acta, 1292(2), 259-264 (1996-02-08)
myo-Inositol monophosphatase catalyzes dephosphorylation of the synthetic substrate anthraniloyl-2'-AMP. Binding of this fluorescent substrate to Tb(III)-monophosphatase was monitored by luminescence spectroscopy. The anthraniloyl chromophore excited at 330 nm sensitizes the long lived luminescence of enzyme bound Tb(III) at 490, 545
William J Rieter et al.
Journal of the American Chemical Society, 130(35), 11584-11585 (2008-08-09)
Pt-containing nanoscale coordination polymer (NCP) particles with the formula of Tb2(DSCP)3(H2O)12 (where DSCP represents disuccinatocisplatin), NCP-1, were precipitated from an aqueous solution of Tb3+ ions and DSCP bridging ligands via the addition of a poor solvent. SEM and TEM images
A Shinohara et al.
Toxicology, 66(1), 93-103 (1991-02-11)
To investigate the biological effects of terbium (Tb), male mice were intraperitoneally administered with TbCl3 at doses of 10, 50, or 250 mg Tb/kg. The Tb distribution in organs was determined after 18-20 h of injection by using a spectrofluorometer.
David L Rosen
Applied optics, 45(13), 3152-3157 (2006-04-28)
An experimental investigation is described of a terbium luminescence sensor for continuous collection and detection of bacterial endospores suspended in the atmosphere. A sensor is presented that continuously monitors and rapidly detects small concentrations of aerosolized bacterial endospores. The aerosol
H Shimada et al.
Journal of toxicology and environmental health, 48(1), 81-92 (1996-05-01)
Terbium (Tb) is a rare earth metal that finds use in several emerging technologies. However, little is known about the biological effects of Tb. Thus, in this study the pulmonary toxicity of systemic Tb in mice was investigated. Mice were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service