325546
Dysprosium(III) chloride
anhydrous, powder, 99.99% trace metals basis
Synonym(s):
Dysprosium trichloride anhydrous, Trichlorodysprosium
About This Item
Recommended Products
grade
anhydrous
Quality Level
Assay
99.99% trace metals basis
form
powder
reaction suitability
reagent type: catalyst
core: dysprosium
impurities
≤150.0 ppm Trace Rare Earth Analysis
mp
680 °C (lit.)
density
3.67 g/mL at 25 °C (lit.)
SMILES string
Cl[Dy](Cl)Cl
InChI
1S/3ClH.Dy/h3*1H;/q;;;+3/p-3
InChI key
BOXVSFHSLKQLNZ-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
accessory
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service