Skip to Content
Merck
All Photos(1)

Documents

451266

Sigma-Aldrich

Scandium(III) chloride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Scandium chloride, Scandium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ScCl3
CAS Number:
Molecular Weight:
151.31
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: scandium

impurities

≤150.0 ppm Trace Rare Earth Analysis

mp

960 °C (lit.)

density

2.39 g/mL at 25 °C (lit.)

SMILES string

Cl[Sc](Cl)Cl

InChI

1S/3ClH.Sc/h3*1H;/q;;;+3/p-3

InChI key

DVMZCYSFPFUKKE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used to form an intermetallic nickel-scandium compound film by potentiostatic cathodic reduction from molten salt mixtures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chem. Abstr., 123, 269239x-269239x (1995)
Tatejwa, K. et al.
Kidorui, 26, 74-74 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service