Skip to Content
Merck
All Photos(2)

Key Documents

450952

Sigma-Aldrich

Lithium iodide

greener alternative

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Lithium monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiI
CAS Number:
Molecular Weight:
133.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
synthesis grade

grade

synthesis grade

Quality Level

product line

AnhydroBeads

Assay

99.999% trace metals basis

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

446 °C (lit.)

density

3.49 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li+].[I-]

InChI

1S/HI.Li/h1H;/q;+1/p-1

InChI key

HSZCZNFXUDYRKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Controls regioselectivity in palladium catalyzed allylic alkylation reactions.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kawatsura, M. et al.
Chemical Communications (Cambridge, England), 217-217 (1998)
Valentina Fiorini et al.
Dalton transactions (Cambridge, England : 2003), 49(41), 14543-14555 (2020-10-14)
A set of three new Ru(ii) polypyridyl complexes decorated with 5-aryl tetrazolato ligands (R-CN4)-, (D series, namely D1, D3 and D4), is presented herein. Whereas complex D1 represents the pyrazinyl tetrazolato analogue of a previously reported Ru(ii) complex (D2) with
Qianglu Lin et al.
Journal of the American Chemical Society, 139(19), 6644-6653 (2017-04-22)
The use of semiconductor nanocrystal quantum dots (QDs) in optoelectronic devices typically requires postsynthetic chemical surface treatments to enhance electronic coupling between QDs and allow for efficient charge transport in QD films. Despite their importance in solar cells and infrared
Yu-il Kang et al.
ChemSusChem, 8(22), 3799-3804 (2015-10-17)
Dye-sensitized solar cells (DSCs) with long-term stability are produced using polymer-gel electrolytes (PGEs). In this study, we introduce the formation of PGEs using in situ gelation with poly(methyl methacrylate) (PMMA) particles and graphene fillers that are pre-deposited on the counter electrodes.
Jianjian Lin et al.
Scientific reports, 4, 5769-5769 (2014-08-30)
Three-dimensional (3D) hierarchical nanoscale architectures comprised of building blocks, with specifically engineered morphologies, are expected to play important roles in the fabrication of 'next generation' microelectronic and optoelectronic devices due to their high surface-to-volume ratio as well as opto-electronic properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service