Merck
  • Phase-Transfer Ligand Exchange of Lead Chalcogenide Quantum Dots for Direct Deposition of Thick, Highly Conductive Films.
Phase-Transfer Ligand Exchange of Lead Chalcogenide Quantum Dots for Direct Deposition of Thick, Highly Conductive Films.
Journal of the American Chemical Society (2017-04-22)
Qianglu Lin, Hyeong Jin Yun, Wenyong Liu, Hyung-Jun Song, Nikolay S Makarov, Oleksandr Isaienko, Tom Nakotte, Gen Chen, Hongmei Luo, Victor I Klimov, Jeffrey M Pietryga
ABSTRACT
The use of semiconductor nanocrystal quantum dots (QDs) in optoelectronic devices typically requires postsynthetic chemical surface treatments to enhance electronic coupling between QDs and allow for efficient charge transport in QD films. Despite their importance in solar cells and infrared (IR) light-emitting diodes and photodetectors, advances in these chemical treatments for lead chalcogenide (PbE; E = S, Se, Te) QDs have lagged behind those of, for instance, II-VI semiconductor QDs. Here, we introduce a method for fast and effective ligand exchange for PbE QDs in solution, resulting in QDs completely passivated by a wide range of small anionic ligands. Due to electrostatic stabilization, these QDs are readily dispersible in polar solvents, in which they form highly concentrated solutions that remain stable for months. QDs of all three Pb chalcogenides retain their photoluminescence, allowing for a detailed study of the effect of the surface ionic double layer on electronic passivation of QD surfaces, which we find can be explained using the hard/soft acid-base theory. Importantly, we prepare highly conductive films of PbS, PbSe, and PbTe QDs by directly casting from solution without further chemical treatment, as determined by field-effect transistor measurements. This method allows for precise control over the surface chemistry, and therefore the transport properties of deposited films. It also permits single-step deposition of films of unprecedented thickness via continuous processing techniques, as we demonstrate by preparing a dense, smooth, 5.3-μm-thick PbSe QD film via doctor-blading. As such, it offers important advantages over laborious layer-by-layer methods for solar cells and photodetectors, while opening the door to new possibilities in ionizing-radiation detectors.
Sigma-Aldrich
Tris(diethylamino)phosphine, 97%
Sigma-Aldrich
Lithium formate monohydrate, 98%
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
2,6-Difluoropyridine, 99%
Sigma-Aldrich
Potassium selenocyanate, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium thiocyanate hydrate
Sigma-Aldrich
Lithium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis